Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252374.1 | NP_001239303.1 | 9 | 680 | ||
NM_026004.3 | NP_080280.3 | 8 | 837 | |||
GenBank mRNA | AK011525.1 | - | 8 | 788 | ||
AK011894.1 | - | 8 | 735 | |||
AK160139.1 | - | 9 | 681 | |||
BC038029.1 | - | 9 | 673 | 4018956 |