Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029153.1 | NP_083429.1 | 7 | 682 | ||
GenBank mRNA | AK015706.1 | - | 6 | 643 | ||
AK132867.1 | - | 6 | 652 | |||
AK135058.1 | - | 7 | 696 | |||
AK154671.1 | - | 7 | 697 | |||
AK162168.1 | - | 7 | 887 | |||
AK162198.1 | - | 7 | 1116 | |||
AK170343.1 | - | 7 | 733 | |||
BC034283.1 | - | 7 | 682 | 4162134 |