Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000607.2 | NP_000598.2 | 3 | 400 | ||
GenBank mRNA | AK312035.1 | - | 3 | 325 | ||
AK316207.1 | - | 2 | 741 | |||
BC026238.1 | - | 3 | 325 | 4767956 | ||
BC104818.1 | - | 3 | 314 | 8143821 | ||
BC104820.1 | - | 3 | 314 | 8143823 | ||
BC143313.1 | - | 3 | 314 | 9051818 | ||
BC143314.1 | - | 3 | 314 | 9051819 | ||
BI908589.1 | - | 3 | 400 | |||
BT019790.1 | - | 3 | 289 | |||
EU794584.1 | - | 3 | 289 | |||
M13692.1 | - | 3 | 304 | |||
X02544.1 | - | 3 | 367 |