Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177328.3 | NP_796302.2 | 8 | 2660 | ||
GenBank mRNA | AK053447.1 | - | 9 | 2710 | ||
BC080315.1 | - | 8 | 2660 | 6853901 |
Set Membership: |
G-protein coupled receptor |