Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028882.4 | NP_083158.3 | 12 | 1795 | ||
GenBank mRNA | AK028900.1 | - | 12 | 1577 | ||
AK031704.1 | - | 12 | 1407 | |||
AK052671.1 | - | 12 | 1796 | |||
AK153839.1 | - | 12 | 1456 | |||
BC138131.1 | - | 12 | 1338 | 8861150 |