Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134081.5 | NP_598842.1 | 2 | 430 | ||
GenBank mRNA | AK043725.1 | - | 2 | 400 | ||
AK075979.1 | - | 2 | 391 | |||
AK077324.1 | - | 2 | 400 | |||
AK153033.1 | - | 2 | 390 | |||
AK162075.1 | - | 2 | 305 | |||
BC014686.1 | - | 2 | 392 | 4481661 | ||
BC023787.1 | - | 2 | 388 | 5346759 | ||
BC027012.1 | - | 2 | 383 | 5367851 | ||
CX238858.1 | - | 2 | 430 |