Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: 2700081O15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175381.6 | NP_780590.3 | 5 | 2901 | ||
GenBank mRNA | AK032941.1 | - | 1 | 1171 | ||
BC059063.1 | - | 3 | 2340 | 6825195 | ||
BC096549.1 | - | 4 | 1940 | |||
BC096638.1 | - | 3 | 2126 | 6825382 |