Gene Symbol: Lhfp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175386.3 | NP_780595.1 | 2 | 765 | ||
GenBank mRNA | AK034532.1 | - | 2 | 603 | ||
AK159126.1 | - | 2 | 754 | |||
AK159549.1 | - | 2 | 764 | |||
BC052079.1 | - | 2 | 765 | 6402782 | ||
BC095934.1 | - | 2 | 609 | 3154659 |