Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmadhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133839.2 | NP_598600.1 | 6 | 609 | ||
GenBank mRNA | BC002253.1 | - | 6 | 609 | 3493526 |