Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam118b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286604.1 | NP_001273533.1 | 5 | 480 | ||
NM_175411.5 | NP_780620.1 | 3 | 350 | |||
NM_194257.2 | NP_919233.1 | 3 | 350 | |||
GenBank mRNA | AK012261.1 | - | 1 | 136 | ||
AK028018.1 | - | 1 | 99 | |||
AK054309.1 | - | 232 | ||||
AK079397.1 | - | 5 | 453 | |||
AK081206.1 | - | 3 | 326 | |||
AK153897.1 | - | 3 | 325 | |||
AK163695.1 | - | 2 | 232 | |||
AK168168.1 | - | 3 | 326 | |||
AK169810.1 | - | 3 | 325 | |||
BC051180.1 | - | 3 | 312 | 6315210 |