Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172778.2 | NP_766366.2 | 7 | 742 | ||
GenBank mRNA | AK031833.1 | - | 7 | 726 | ||
AK054050.1 | - | 7 | 741 | |||
BC113182.1 | - | 7 | 731 | 40061991 | ||
BC113788.1 | - | 7 | 731 | 40061990 |