Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Art3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181728.2 | NP_859417.2 | 4 | 937 | ||
GenBank mRNA | AJ311769.1 | - | 2 | 721 | ||
AJ311770.1 | - | 2 | 721 | |||
AJ311771.1 | - | 2 | 721 | |||
AJ311772.1 | - | 2 | 721 | |||
AJ311773.1 | - | 2 | 721 | |||
AK016257.1 | - | 4 | 930 | |||
BC141069.1 | - | 2 | 797 | 9055681 | ||
BC145327.1 | - | 2 | 797 | 9053883 | ||
BE625316.1 | - | 1 | 62 |