Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phka2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177878.1 | NP_001171349.1 | 21 | 2702 | ||
NM_001177879.1 | NP_001171350.1 | 22 | 2516 | |||
NM_172783.3 | NP_766371.1 | 21 | 2702 | |||
GenBank mRNA | AK052346.1 | - | 21 | 2447 | ||
AK052992.1 | - | 21 | 2703 | |||
AK165506.1 | - | 22 | 2517 | |||
BC050040.1 | - | 22 | 2475 | 6431743 | ||
BC051167.1 | - | 16 | 1944 |