Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phka2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177878.1 | NP_001171349.1 | 5 | 899 | ||
NM_001177879.1 | NP_001171350.1 | 6 | 713 | |||
NM_172783.3 | NP_766371.1 | 5 | 899 | |||
GenBank mRNA | AK052346.1 | - | 5 | 644 | ||
AK052992.1 | - | 5 | 900 | |||
AK165506.1 | - | 6 | 714 | |||
BC050040.1 | - | 6 | 672 | 6431743 | ||
BC058254.1 | - | 5 | 868 |