Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fdps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253751.1 | NP_001240680.1 | 7, 8 | 930 | ||
NM_134469.4 | NP_608219.1 | 6, 7 | 714 | |||
GenBank mRNA | AF309508.1 | - | 5, 6 | 619 | ||
AK088601.1 | - | 6, 7 | 634 | |||
AK140881.1 | - | 6, 7 | 706 | |||
AK151742.1 | - | 6, 7 | 643 | |||
AK166026.1 | - | 6, 7 | 633 | |||
AK168008.1 | - | 6, 7 | 633 | |||
BC048497.1 | - | 6, 7 | 692 | 6531586 | ||
BC087886.1 | - | 6, 7 | 637 | 6595030 | ||
CT010365.1 | - | 5, 6 | 586 |