Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgkg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138650.2 | NP_619591.1 | 19 | 2073 | ||
GenBank mRNA | AK083258.1 | - | 18 | 1949 | ||
AK087972.1 | - | 19 | 2284 | |||
AK220294.1 | - | 19 | 2015 | |||
BC015278.1 | - | 18 | 1646 | 4221290 |