Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026662.4 | NP_080938.1 | 6 | 942 | ||
GenBank mRNA | AK012819.1 | - | 6 | 974 | ||
AK019169.2 | - | 6 | 911 | |||
AK029690.1 | - | 6 | 957 | |||
AK045007.1 | - | 6 | 943 | |||
BC024942.1 | - | 6 | 929 | 4022452 |