Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163529.1 | NP_001157001.1 | 17 | 2222 | ||
NM_033615.2 | NP_291093.2 | 18 | 2300 | |||
GenBank mRNA | AB059632.1 | - | 18 | 2300 | ||
AB059633.1 | - | 17 | 2222 | |||
AF386072.1 | - | 14 | 1667 | |||
AF472524.1 | - | 18 | 2032 | |||
AK048367.1 | - | 13 | 1656 | |||
AK053825.1 | - | 14 | 2719 | |||
AY054984.1 | - | 18 | 2000 | |||
AY382193.1 | - | 17 | 1953 | |||
AY382194.1 | - | 18 | 2031 | |||
BC138139.1 | - | 17 | 1945 | 8861158 |