Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177084.3 | NP_796058.1 | 11 | 2252 | ||
GenBank mRNA | AK085681.1 | - | 11 | 2278 | ||
AK141534.1 | - | 11 | 2253 | |||
AK170522.1 | - | 9 | 1091 | |||
BC120543.1 | - | 11 | 2170 | 40129466 |