Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030201.3 | NP_084477.1 | 5 | 1084 | ||
GenBank mRNA | AK030553.1 | - | 6 | 1153 | ||
AK034643.1 | - | 5 | 1064 | |||
BC039701.1 | - | 4 | 1025 | |||
BC085181.1 | - | 5 | 1044 | 30646642 | ||
BC094508.1 | - | 6 | 1123 |