Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053205.1 | NP_444435.1 | 1 | 666 | ||
GenBank mRNA | BC125368.1 | - | 1 | 716 | 40129883 | |
BC125370.1 | - | 1 | 716 | 40129885 |
Set Membership: |
![]() |