Whole Allergen
k76 Isocyanate MDI
k76 Isocyanate MDI Scientific Information
Type:
Whole Allergen
Display Name:
Isocyanate MDI
Route of Exposure
Main
Isocyanates are widely used in the production of polyurethane plastics, glues and paints. Individuals at risk are polyurethane foam makers, upholsterers, spray painters, wire coating workers, plastic molders, rubber workers, fabric coaters, metal rod coaters, truck body insulators and persons applying artificial turf and others.
Unexpected exposure
Isocyanates have been mentioned in relation to the Sick Building Syndrome.
Environment
Any workplace where isocyanates are used in significant amounts.
Clinical Relevance
IgE-mediated reactions
A wide spectrum of potentially allergic respiratory symptoms have been described and up to 5% of exposed workers have been reported to have developed asthma at work (1). IgE antibodies to isocyanates have been found in 14-19% of sensitised workers (2,3).
Other reactions
Non-immunological, irritative mechanisms are likely to be of importance
Molecular Aspects
Cross-reactivity
The three isocyanates (TDI, MDI and HDI) are closely related structurally being HSA-conjugated haptens and there is some allergenic crossreactivity (2). However, the major specificity depends on the type of isocyanate exposure.
Compiled By
Last Reviewed : June 2022
References
  1. Butcher BT, Karr RM, O’Neil CE, Wilson MR, Dharmarajan V, Salvaggio JE, Weill H: Inhalation challenge and pharmacologic studies of toluene di-isocyanate(TDI)-sensitive workers. J Allergy Clin Immunol 1979, 64, 146-152
  2. Baur X: Immunologic cross-reactivity between different albumin-bound isocyanates. J Allergy Clin Immunol 1983, 71, 197-205..
  3. Butcher BT, O’Neil CE, Reed MA, Salvaggio JE: Radioallergosorbent testing of toluene di-isocyanate-reactive individuals using p-tolyl isocyanate antigen. J Allergy Clin Immunl 1980, 66, 213-216.