Mouse fibroblasts. BODIPY® FL phallacidin, Texas Red® goat anti–mouse IgG (H+L) antibody and DAPI.
Photomicrograph of mouse fibroblasts that have been formaldehyde-fixed, acetone-permeabilized and triple-stained with the F-actin–specific probe BODIPY® FL phallacidin (Cat. no. B607), with mouse monoclonal anti–ß-tubulin antibody in conjunction with Texas Red® goat anti–mouse IgG antibody (Cat. no. T862) and with DAPI (Cat. no. D1306, D3571, D21490). The image was obtained by taking multiple exposures through bandpass optical filter sets appropriate for fluorescein, Texas Red® dye and DAPI using a 100X Plan Apochromat objective.
Photomicrograph of mouse fibroblasts that have been formaldehyde-fixed, acetone-permeabilized and triple-stained with the F-actin–specific probe BODIPY® FL phallacidin (Cat. no. B607), with mouse monoclonal anti–ß-tubulin antibody in conjunction with Texas Red® goat anti–mouse IgG antibody (Cat. no. T862) and with DAPI (Cat. no. D1306, D3571, D21490).  The image was obtained by taking multiple exposures through bandpass optical filter sets appropriate for fluorescein, Texas Red® dye and DAPI using a 100X Plan Apochromat objective.
Related Products
Catalog # Name Size Price (USD) Qty
D1306 DAPI (4',6-diamidino-2-phenylindole, dihydrochloride), 10 mg, 10 mg Each
152.65
Online Exclusive
156.00
Save 3.35 (2%)
D3571 DAPI (4',6-diamidino-2-phenylindole, dilactate), 10 mg, 10 mg Each
151.65
Online Exclusive
156.00
Save 4.35 (3%)
Related Images