Search Thermo Fisher Scientific
Murashige and Skoog Salt and Vitamin Mixture (1) is a complete medium for plant tissue culture which contains both basal salts and essential vitamins in a premixed format for convenience and time savings. This mixture is simply dissolved in the appropriate volume of water and autoclaved. For semi-solid culture systems agar is added before autoclaving.
Catalog Number(s)
Components | Molecular Weight | Concentration (mg/L) | mM |
---|---|---|---|
Vitamins | |||
Myo-Inositol | 180.0 | 100.0 | 0.5555556 |
Nicotinic acid (Niacin) | 123.0 | 0.05 | 4.0650408E-4 |
Pyridoxine hydrochloride | 206.0 | 0.05 | 2.4271845E-4 |
Thiamine hydrochloride | 337.0 | 0.1 | 2.967359E-4 |
Inorganic Salts | |||
Ammonium Nitrate (NH4NO3) | 80.0 | 1650.0 | 20.625 |
Boric Acid (H3BO3) | 62.0 | 6.2 | 0.099999994 |
Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 332.2 | 2.9927928 |
Cobalt Chloride (CoCl2-6H2O) | 238.0 | 0.025 | 1.0504202E-4 |
Cupric Sulfate (CuSO4-6H20) | 250.0 | 0.025 | 1.00000005E-4 |
Ferric sulfate (FeSO4-7H2O) | 278.0 | 27.8 | 0.099999994 |
Magnesium Sulfate (MgSO4) (anhyd.) | 120.0 | 180.7 | 1.5058333 |
Manganese Sulfate (MnSO4-H20) | 169.0 | 16.9 | 0.099999994 |
Potassium Iodide (KI) | 166.0 | 0.83 | 0.005 |
Potassium Nitrate (KNO3) | 101.0 | 1900.0 | 18.811882 |
Potassium Phosphate monobasic (KH2PO4) | 136.0 | 170.0 | 1.25 |
Sodium Molybdate (Na2MoO4-2H20) | 243.0 | 0.25 | 0.0010288066 |
Zinc sulfate (ZnSO4-7H2O) | 288.0 | 8.6 | 0.029861113 |
Other Components | |||
Glycine (Free Base) | 75.0 | 2.0 | 0.026666667 |
Sodium EDTA (Na2-EDTA) | 372.0 | 37.26 | 0.100161284 |
1. Murashige, T. and Skoog, F. (1962) Physiol. Plant. 15, 485.