Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This peptide corresponds to 16 amino acids near the amino terminus of human CCDC69A.
PEP-1049 can be used as a blocking peptide with polyclonal antibody PA5-20935.
May act as a scaffold to regulate the recruitment and assembly of spindle midzone components. Required for the localization of AURKB and PLK1 to the spindle midzone. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coiled-coil domain-containing protein 69
Gene Aliases: CCDC69
UniProt ID: (Human) A6NI79
Entrez Gene ID: (Human) 26112
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support