Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-145224. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The FYVE domain mediates the recruitment of proteins involved in membrane trafficking and cell signaling to phosphatidylinositol 3-phosphate (PtdIns(3)P)-containing membranes. This gene encodes a protein which contains a single FYVE domain and multiple WD40 repeats. This protein is localized to endosomes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FENS-1; FYVE domain-containing protein localized to endosomes 1; Phosphoinositide-binding protein 1; phosphoinositide-binding protein SR1; WD repeat and FYVE domain-containing protein 1; WD40- and FYVE domain-containing protein 1; Zinc finger FYVE domain-containing protein 17
Gene Aliases: FENS-1; FENS1; KIAA1435; WDF1; WDFY1; ZFYVE17
UniProt ID: (Human) Q8IWB7
Entrez Gene ID: (Human) 57590
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support