Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Proteins & Peptides
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Proteins & Peptides ›
            • CD32b (FCGR2B) Proteins

            Invitrogen

            Human Fc gamma RIIB (CD32b) His-tag Recombinant Protein

            View all (2) CD32b (FCGR2B) proteins
            Datasheet
            Tech Support
            Catalog # A42520
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            367.00
             
            Check your price
            Request bulk or custom format
            Datasheet
            Tech Support

            Cite Human Fc gamma RIIB (CD32b) His-tag Recombinant Protein

            Product Details

            A42520

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            Assay-dependent
            -

            Immunohistochemistry (IHC)

            Assay-dependent
            -

            ELISA (ELISA)

            Assay-dependent
            -
            Product Specifications

            Species

            Human

            Expression System

            HEK293 cells

            Amino acid sequence

            Human CD32b, amino acids Ala46-Pro217 (Accession # NP_001002274.1) with an N-terminal signal peptide and a C-terminal His-tag

            Tag

            His-tag

            Molecular weight

            20.8 kDa

            Class

            Recombinant

            Type

            Protein

            Purity

            ≥95 % as determined by SDS-PAGE. ≥95 % as determined by SEC-HPLC.

            Endotoxin concentration

            <1.0 EU/µg

            Activity

            Loaded Recombinant Human CD32B/Fcgr2 Protein, His Tag on His1K Biosensor, can bind Rituximab with an affinity constant of 1.13 µM as determined in BLI assay.

            Conjugate

            Unconjugated

            Form

            Lyophilized

            Purification

            purified

            Storage buffer

            PBS, pH 7.4, with 5% mannitol, 5% trehalose, 0.01% Tween 80

            Contains

            no preservative

            Storage conditions

            -20°C or -80°C if preferred

            Shipping conditions

            Ambient

            Product Specific Information

            Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/mL or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

            Target Information

            This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: CD32; CD32 receptor 2; CD32 receptor 2a; CD32 receptor 2b; CD32A; CDw32; Fc fragment of IgG, low affinity II, receptor for (CD32); Fc fragment of IgG, low affinity IIb, receptor (CD32); Fc fragment of IgG, low affinity IIb, receptor for (CD32); Fc gamma receptor IIb; Fc gamma RIIb; fc-gamma RII-a; Fc-gamma RII-b; fc-gamma-RIIa; Fc-gamma-RIIb; FcGR; FCGR2A1; fcRII-a; fcRII-b; FGFR2B; IgG Fc receptor II-b; Low affinity immunoglobulin gamma Fc region receptor II-b

            View more View less

            Gene Aliases: CD32; CD32B; FCG2; FCGR2; FCGR2B; IGFR2

            View more View less

            UniProt ID: (Human) P31994

            View more View less

            Entrez Gene ID: (Human) 2213

            View more View less

            It has to be done as per old AB suggested Products section.
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-6ff95d844f-6cgcf:80/100.66.79.97:80.
            git-commit: f44aa9153aef922f8fad9a05e87b8aee811da128
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.28.0-Offline