Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Description: Human IL-10 is a ~18 kDa factor also known as cytokine synthesis inhibitory factor (CSIF). Th2 T cells, B1 B cells and keratinocytes are the major cell subsets that produce IL-10. IL-10 inhibits synthesis of Th1 cytokines and proliferation of T cells and acts as a costimulatory signal for mast cells, developing thymocytes and Th2 response.
Applications Reported: Human IL-10 RSG ELISA Lyophilized Standard has been reported for use in the Human IL-10 ELISA RSG (Product # 88-7106).
Applications Tested: The intended use of the Human IL-10 RSG ELISA Lyophilized Standard is as a standard for the Human IL-10 ELISA (Product # 88-7106-77). Typically the standard is run in a series of 7 doubling dilutions from the 'top' standard. Please refer to the Certificate of Analysis included with each ELISA set for instructions on the proper reconstitution and dilution of the lyophilized standard.
Note: These are single-use vials. Use one time within 4 hours of reconstitution and discard. Reconstitute and dilute as suggested in the Certificate of Analysis or recovery may not be accurate.
Source: E. coli expressed amino acids Ser 19-Asn 178 of mature human IL-10 accession # NM_000572.
IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CSIF; Cytokine synthesis inhibitory factor; H-IL-10; il 10; IL-10; ILN; Interleukin; Interleukin-10; Interleukin10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor
Gene Aliases: CSIF; GVHDS; IL-10; IL10; IL10A; TGIF
UniProt ID: (Human) P22301
Entrez Gene ID: (Human) 3586
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support