Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
It is recommended that the protein be aliquoted for optimal storage.
It is recommended that sterile water (848 µL) be added to the vial to prepare a stock solution of 1.18 mg/mL.
Concentration is measured by UV.
LILRA3 is a member of leukocyte immunoglobulin-like receptor gene family. Leukocyte Ig-like receptors (LIRs) are a family of inhibitory and stimulatory immunoreceptors expressed predominantly on monocytes and B cells and at lower levels on dendritic cells and natural killer (NK) cells. LILRA3 lacks a transmembrane region and may act as a soluble receptor for class I MHC antigens. It is expressed in B-cells, and at lower levels in natural killer (NK) cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD85 antigen-like family member I; CD85i; leucocyte Ig-like receptor A1; Leukocyte immunoglobulin-like receptor 6; Leukocyte immunoglobulin-like receptor subfamily A member 1; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 1; LIR-6
Gene Aliases: CD85I; LILRA1; LIR-6; LIR6
UniProt ID: (Human) O75019
Entrez Gene ID: (Human) 11024
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support