Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
It is recommended that the protein be aliquoted for optimal storage.
It is recommended that sterile water (1205 µL) be added to the vial to prepare a stock solution of 0.83 mg/mL.
Concentration is measured by UV.
Leukocyte Ig-like receptors (LIRs) are a family of immunoreceptors expressed predominantly on monocytes and B cells and at lower levels on dendritic cells and natural killer (NK) cells. All LIRs in subfamily B have an inhibitory function (LILRB1; MIM 604811). LIRs in subfamily A, with short cytoplasmic domains lacking an immunoreceptor tyrosine-based inhibitory motif (ITIM) and with trans membrane regions containing a charged arginine residue, may initiate stimulatory cascades. One member of subfamily A (LILRA3; MIM 604818) lacks a transmembrane region and is presumed to be a soluble receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD85 antigen-like family member H; CD85h; ILT-1; Immunoglobulin-like transcript 1; leucocyte Ig-like receptor A2; Leukocyte immunoglobulin-like receptor 7; Leukocyte immunoglobulin-like receptor subfamily A member 2; leukocyte immunoglobulin-like receptor subfamily A member 2 soluble; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 2; LIR-7
Gene Aliases: CD85H; ILT1; LILRA2; LIR-7; LIR7
UniProt ID: (Human) Q8N149
Entrez Gene ID: (Human) 11027
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support