Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%).
This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
All cells within tissues are surrounded by an extracellular matrix (ECM) giving the tissues shape and structure. The ECM is constantly being remodeled and constant communication is maintained between cells through this matrix. Secreted proteins, termed matrix metalloproteinases (MMPs), are involved in the modulation of cell-matrix interactions. MMPs are Zn (2+)-binding endopeptidases that degrade various components of the ECM. MMPs are enzymes implicated in normal and pathologic tissue remodeling processes, wound healing, angiogenesis, and tumor invasion. These enzymes are very potent when active, and are associated with extracellular space inhibitors called TIMPs (tissue inhibitors of matrix metalloproteinases). TIMPs have been shown to block tumor cell invasion suggesting that they act as metastasis suppressor genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: matrix metallo protease; matrix metallopeptidase 24 (membrane-inserted); matrix metalloproteinase 24 (membrane-inserted); Matrix metalloproteinase-24; membrane-type 5 matrix metalloproteinase; Membrane-type matrix metalloproteinase 5; Membrane-type-5 matrix metalloproteinase; MMP; MMP-24; MMPs; MT-MMP 5; MT5-MMP; RP4-614O4.10
Gene Aliases: MMP-24; MMP24; MMP25; MT-MMP 5; MT-MMP5; MT5-MMP; MT5MMP; MTMMP5
UniProt ID: (Human) Q9Y5R2
Entrez Gene ID: (Human) 10893
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support