Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Proteins & Peptides
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Proteins & Peptides ›
            • GDF11 Proteins
            The PeproTech website has now moved to Thermofisher.com. All orders for PeproTech products must be placed on thermofisher.com or offline via phone, fax, email, or distributor partner. For any questions, please contact PeproTech.CustomerService@thermofisher.com

            Gibco

            Human/Mouse/Rat GDF-11 Recombinant Protein, PeproTech®

            24 References
            View all (3) GDF11 proteins
            Datasheet
            Tech Support
            Promotion Available
            Learn more
            Catalog # 120-11-20UG
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            212.40
            Special offer
            Online exclusive
            Ends: 16-Jun-2025
            236.00
            Save 23.60 (10%)
             
            In stock
            Check your price
            Request bulk or custom format
            Datasheet
            Tech Support

            Cite Human/Mouse/Rat GDF-11 Recombinant Protein, PeproTech®

            • Testing Data (2)
            Application
            Human/Mouse/Rat GDF-11 Protein in Functional Assay (Functional)
            Group 53 Created with Sketch.

            FIGURE: 1 / 2

            Human/Mouse/Rat GDF-11 Protein (120-11-20UG) in Functional

            Bioassay analysis of Human/Mouse/Rat GDF-11 Recombinant Protein, PeproTech® (Product # 120-11-1MG).
            View Product

            Human/Mouse/Rat GDF-11 Protein in Functional Assay (Functional)
            Human/Mouse/Rat GDF-11 Protein in Functional Assay (Functional)

            Product Details

            120-11-20UG

            Applications
            Tested Dilution
            Publications

            Functional Assay (Functional)

            Assay-dependent
            View 2 publications 2 publications

            Dot blot (DB)

            -
            View 1 publication 1 publication

            In vitro Assay (IV)

            -
            View 10 publications 10 publications

            Miscellaneous PubMed (Misc)

            -
            View 11 publications 11 publications
            Product Specifications

            Species

            Human, Mouse, Rat

            Published species

            Human, Mouse, Rat

            Expression System

            E. coli

            Amino acid sequence

            NLGLDCDEHS SESRCCRYPL TVDFEAFGWD WIIAPKRYKA NYCSGQCEYM FMQKYPHTHL VQQANPRGSA GPCCTPTKMS PINMLYFNDK QQIIYGKIPG MVVDRCGCS

            Molecular weight

            25 kDa

            Class

            Recombinant

            Type

            Protein

            Purity

            ≥ 98% by SDS-PAGE gel and HPLC analyses.

            Endotoxin concentration

            <1 EU/µg

            Activity

            Assay 1: Determined by its ability to inhibit induced alkaline phosphatase production by ATDC-5 chondrogenic cells. The expected ED50 for this effect is 0.08-0.10 ug/ml. Assay 2: Determined by its ability to inhibit alkaline phosphatase activity in differentiating MC3T3/E1cells. The ED50 for this effect is 8-10 ng/ml.

            Conjugate

            Unconjugated

            Form

            Lyophilized

            Purification

            purified

            Contains

            no preservative

            Storage conditions

            -20°C

            Shipping conditions

            Ambient

            Product Specific Information

            120-11-1MG will be provided as 2 x 500 µg (120-11-500UG).

            Recombinant Human/Murine/Rat GDF-11 is a 25.0 kDa disulfide-linked homodimer containing two 109 amino acid polypeptide chains. It is highly homologous to myostatin/GDF-8, sharing 90% amino acid sequence identity.

            This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

            Target Information

            GDF11 is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in mice and Xenopus suggest that this protein is involved in mesodermal formation and neurogenesis during embryonic development.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: BMP-11; Bone morphogenetic protein 11; GDF-11; Growth/differentiation factor 11

            View more View less

            Gene Aliases: BMP-11; BMP11; GDF11

            View more View less

            UniProt ID: (Human) O95390, (Mouse) Q9Z1W4, (Rat) Q9Z217

            View more View less

            Entrez Gene ID: (Human) 10220, (Mouse) 14561, (Rat) 29454

            View more View less

            It has to be done as per old AB suggested Products section.
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-6l4vp:80/100.66.76.166:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0