Product References
The adipose-neural axis is involved in epicardial adipose tissue-related cardiac arrhythmias.
Cell reports. Medicine
Fan Y,Huang S,Li S,Wu B,Zhao Q,Huang L,Zheng Z,Xie X,Liu J,Huang W,Sun J,Zhu X,Zhu J,Xiang AP,Li W
450-03 was used in In vitro experiments to show that adipocyte-derived leptin activates sympathetic neurons and increases the release of neuropeptide Y (NPY), which in turn triggers arrhythmia in cardiomyocytes by interacting with the Y1 receptor (Y1R) and subsequently enhancing the activity of the Na+/Ca2+ exchanger (NCX) and calcium/calmodulin-dependent protein kinase II (CaMKII).
Tue May 21 00:00:00 UTC 2024
Apolipoprotein E ε4 disrupts oligodendrocyte differentiation by interfering with astrocyte-derived lipid transport.
Journal of neurochemistry
Mok KK,Yeung SH,Cheng GW,Ma IW,Lee RH,Herrup K,Tse KH
450-03 was used in In vitro experiments to propose that high myelin turnover and OL exhaustion found in APOE4 carriers is a likely explanation for the APOE-dependent myelin phenotypes of the AD brain.
Sat Apr 01 00:00:00 UTC 2023
Efficient generation of functional neurons from mouse embryonic stem cells via neurogenin-2 expression.
Nature protocols
Liu Y,Wang J,Südhof TC,Wernig M
450-03 was used in Cell Culture to provide a detailed approach for the efficient and reproducible generation of functional iN cells from mouse ESC cultures by the genetically induced expression of neurogenin-2.
Sun Oct 01 00:00:00 UTC 2023
An engineered Sox17 induces somatic to neural stem cell fate transitions independently from pluripotency reprogramming.
Science advances
Weng M,Hu H,Graus MS,Tan DS,Gao Y,Ren S,Ho DHH,Langer J,Holzner M,Huang Y,Ling GS,Lai CSW,Francois M,Jauch R
450-03 was used in In vitro experiments to report iNSC reprogramming from embryonic and aged mouse fibroblasts as well as from human blood using an engineered Sox17 (eSox17FNV).
Fri Aug 25 00:00:00 UTC 2023
Temporal morphogen gradient-driven neural induction shapes single expanded neuroepithelium brain organoids with enhanced cortical identity.
Nature communications
Pagliaro A,Finger R,Zoutendijk I,Bunschuh S,Clevers H,Hendriks D,Artegiani B
450-03 was used in Cell Culture to find that a prolonged, decreasing gradient of TGF-β signaling is a determining factor in expanded neuroepithelium organoid formation and allows for an extended phase of neuroepithelium expansion.
Tue Nov 28 00:00:00 UTC 2023
Differential response of C9orf72 transcripts following neuronal depolarization.
iScience
Ghaffari LT,Trotti D,Haeusler AR
450-03 was used in Cell Culture to reveal the impact of depolarization on C9orf72 transcripts, and how this response diverges in C9-NRE-carriers, which may have important implications in the underlying unique clinical associations of C9-NRE transcripts and disease pathogenesis.
Fri Jun 16 00:00:00 UTC 2023
Differentiation state and culture conditions impact neural stem/progenitor cell-derived extracellular vesicle bioactivity.
Biomaterials science
Dutta D,Pirolli NH,Levy D,Tsao J,Seecharan N,Wang Z,Xu X,Jia X,Jay SM
450-03 was used in In vitro experiments to assess whether oligodendrocyte precursor cells and immature oligodendrocytes, which are further differentiated than neural progenitor/stem cells (NPSCs) and which both give rise to mature myelinating oligodendrocytes, could yield extracellular vesicles with neurotherapeutic properties comparable or superior to those from NPSCs.
Tue Aug 08 00:00:00 UTC 2023
Higher throughput workflow with sensitive, reliable and automatic quantification of myelination in vitro suitable for drug screening.
Scientific reports
Seiler S,Wälti CM,de Barros V,Barbash S,Foo LC
450-03 was used in Cell Culture to develop a method allowing systematic and reliable testing of remyelinating compound efficacy.
Sat Feb 18 00:00:00 UTC 2023
Multiple genes in a single GWAS risk locus synergistically mediate aberrant synaptic development and function in human neurons.
Cell genomics
Zhang S,Zhang H,Forrest MP,Zhou Y,Sun X,Bagchi VA,Kozlova A,Santos MD,Piguel NH,Dionisio LE,Sanders AR,Pang ZP,He X,Penzes P,Duan J
450-03 was used in Cell Culture to reveal that multiple genes at a single GWAS risk locus mediate a compound effect on neural function, providing a mechanistic link between a non-coding risk variant and disease-related cellular phenotypes.
Wed Sep 13 00:00:00 UTC 2023
Glioma synapses recruit mechanisms of adaptive plasticity.
Nature
Taylor KR,Barron T,Hui A,Spitzer A,Yalçin B,Ivec AE,Geraghty AC,Hartmann GG,Arzt M,Gillespie SM,Kim YS,Maleki Jahan S,Zhang H,Shamardani K,Su M,Ni L,Du PP,Woo PJ,Silva-Torres A,Venkatesh HS,Mancusi R,Ponnuswami A,Mulinyawe S,Keough MB,Chau I,Aziz-Bose R,Tirosh I,Suvà ML,Monje M
450-03 was used in Cell Culture to indicate that brain-derived neurotrophic factor1-3-TrkB signalling promotes malignant synaptic plasticity and augments tumour progression.
Wed Nov 01 00:00:00 UTC 2023
C9orf72 poly(PR) mediated neurodegeneration is associated with nucleolar stress.
iScience
Cicardi ME,Hallgren JH,Mawrie D,Krishnamurthy K,Markandaiah SS,Nelson AT,Kankate V,Anderson EN,Pasinelli P,Pandey UB,Eischen CM,Trotti D
450-03 was used in Cell Culture to find evidence for the induction of programmed cell death via caspase-3 activation.
Fri Sep 15 00:00:00 UTC 2023
Assembloid CRISPR screens reveal impact of disease genes in human neurodevelopment.
Nature
Meng X,Yao D,Imaizumi K,Chen X,Kelley KW,Reis N,Thete MV,Arjun McKinney A,Kulkarni S,Panagiotakos G,Bassik MC,Pașca SP
450-03 was used in In vitro experiments to highlight the power of this CRISPR-assembloid platform to systematically map NDD genes onto human development and reveal disease mechanisms.
Sun Oct 01 00:00:00 UTC 2023
Protocol Optimization for Direct Reprogramming of Primary Human Fibroblast into Induced Striatal Neurons.
International journal of molecular sciences
Kraskovskaya N,Bolshakova A,Khotin M,Bezprozvanny I,Mikhailova N
450-03 was used in Cell Culture to demonstrate that iSNs are functionally active and able to form synaptic connections in co-cultures with mouse cortical neurons.
Wed Apr 05 00:00:00 UTC 2023
Transcriptomics of Hirschsprung disease patient-derived enteric neural crest cells reveals a role for oxidative phosphorylation.
Nature communications
Li Z,Lui KN,Lau ST,Lai FP,Li P,Chung PH,Wong KK,Tam PK,Garica-Barcelo MM,Hui CC,Sham PC,Ngan ES
450-03 was used in In vitro experiments to define various factors associated with Hirschsprung pathogenesis and demonstrate the implications of oxidative phosphorylation in enteric neural crest development.
Sat Apr 15 00:00:00 UTC 2023
Accelerated differentiation of human induced pluripotent stem cells into regionally specific dorsal and ventral spinal neural progenitor cells for application in spinal cord therapeutics.
Frontiers in neuroscience
Huntemer-Silveira A,Malone D,Frie A,Walsh P,Parr AM
450-03 was used in In vitro experiments to describe here the production of a diverse population of both ventral spinal and dorsal horn progenitor cells.
Sat Feb 10 00:00:00 UTC 2024
Extracellular Vesicle Treatment Alleviates Neurodevelopmental and Neurodegenerative Pathology in Cortical Spheroid Model of Down Syndrome.
International journal of molecular sciences
Campbell NB,Patel Y,Moore TL,Medalla M,Zeldich E
450-03 was used in Cell Culture to show the efficacy of EVs in mitigating DS and AD-related cellular phenotypes and pathological depositions in human CS.
Thu Feb 09 00:00:00 UTC 2023
Boosting peripheral BDNF rescues impaired in vivo axonal transport in CMT2D mice.
JCI insight
Sleigh JN,Villarroel-Campos D,Surana S,Wickenden T,Tong Y,Simkin RL,Vargas JNS,Rhymes ER,Tosolini AP,West SJ,Zhang Q,Yang XL,Schiavo G
450-03 was used in In vivo experiments to suggest that selectively targeting muscles with BDNF-boosting therapies could represent a viable therapeutic strategy for CMT2D.
Mon May 08 00:00:00 UTC 2023
Human iPS cell-derived sensory neurons can be infected by SARS-CoV-2.
iScience
Flamier A,Bisht P,Richards A,Tomasello DL,Jaenisch R
450-03 was used in Cell Culture to indicate that sensory neurons can be infected by the original WA1/2020 strain of SARS-CoV-2 as well as the delta and omicron variants, yet infectability differs between the original strain and the variants.
Fri Sep 15 00:00:00 UTC 2023
Three-dimensional co-culture platform of human induced pluripotent stem cell-derived oligodendrocyte lineage cells and neurons for studying myelination.
STAR protocols
von der Bey M,De Cicco S,Zach S,Hengerer B,Ercan-Herbst E
450-03 was used in In vitro experiments to describe steps to differentiate hiPSCs into cortical neurons and oligodendrocyte lineage cells on 3D nanofibers.
Fri Mar 17 00:00:00 UTC 2023
MHC class I and MHC class II reporter mice enable analysis of immune oligodendroglia in mouse models of multiple sclerosis.
eLife
Harrington EP,Catenacci RB,Smith MD,Heo D,Miller CE,Meyers KR,Glatzer J,Bergles DE,Calabresi PA
450-03 was used in Cell Culture to define oligodendrocytes and their progenitors in response to inflammatory demyelination, providing a means to monitor MHC activation in diverse cell types in living mice and define their roles in aging, injury, and disease.
Fri Apr 14 00:00:00 UTC 2023
A CRISPRi/a platform in human iPSC-derived microglia uncovers regulators of disease states.
Nature neuroscience
Dräger NM,Sattler SM,Huang CT,Teter OM,Leng K,Hashemi SH,Hong J,Aviles G,Clelland CD,Zhan L,Udeochu JC,Kodama L,Singleton AB,Nalls MA,Ichida J,Ward ME,Faghri F,Gan L,Kampmann M
450-03 was used in Cell Culture to present a screening platform to systematically elucidate functional consequences of genetic perturbations in human induced pluripotent stem cell-derived microglia.
Thu Sep 01 00:00:00 UTC 2022
Long-term corneal recovery by simultaneous delivery of hPSC-derived corneal endothelial precursors and nicotinamide.
The Journal of clinical investigation
Li Z,Duan H,Jia Y,Zhao C,Li W,Wang X,Gong Y,Dong C,Ma B,Dou S,Zhang B,Li D,Cao Y,Xie L,Zhou Q,Shi W
Tue Jan 04 00:00:00 UTC 2022
Heterozygous expression of the Alzheimer's disease-protective PLCγ2 P522R variant enhances Aβ clearance while preserving synapses.
Cellular and molecular life sciences : CMLS
Solomon S,Sampathkumar NK,Carre I,Mondal M,Chennell G,Vernon AC,Ruepp MD,Mitchell JC
450-03 was used in Sample Preparation to investigate P522R in Alzheimer’s disease.
Wed Jul 27 00:00:00 UTC 2022
Recapitulation of endogenous 4R tau expression and formation of insoluble tau in directly reprogrammed human neurons.
Cell stem cell
Capano LS,Sato C,Ficulle E,Yu A,Horie K,Kwon JS,Burbach KF,Barthélemy NR,Fox SG,Karch CM,Bateman RJ,Houlden H,Morimoto RI,Holtzman DM,Duff KE,Yoo AS
450-03 was used in Cell Culture to demonstrate the utility of miRNA-induced neuronal reprogramming to recapitulate endogenous tau regulation comparable with the adult brain in health and disease.
Thu Jun 02 00:00:00 UTC 2022
Coactivator-associated arginine methyltransferase 1 controls oligodendrocyte differentiation in the corpus callosum during early brain development.
Developmental neurobiology
Ishino Y,Shimizu S,Tohyama M,Miyata S
450-03 was used in Cell Culture to propose that Carm1 is an essential molecule for the development of OPCs and OLs during brain development.
Fri Apr 01 00:00:00 UTC 2022
The m6A reader YTHDF2 is a negative regulator for dendrite development and maintenance of retinal ganglion cells.
eLife
Niu F,Han P,Zhang J,She Y,Yang L,Yu J,Zhuang M,Tang K,Shi Y,Yang B,Liu C,Peng B,Ji SJ
450-03 was used in Cell Culture to show that the N6-methyladenosine (m6A) reader YTHDF2 is highly expressed in mouse retinal ganglion cells.
Fri Feb 18 00:00:00 UTC 2022
OCT4-induced oligodendrocyte progenitor cells promote remyelination and ameliorate disease.
NPJ Regenerative medicine
Yun W,Choi KA,Hwang I,Zheng J,Park M,Hong W,Jang AY,Kim JH,Choi W,Kim DS,Kim IY,Kim YJ,Liu Y,Yoon BS,Park G,Song G,Hong S,You S
Thu Jan 13 00:00:00 UTC 2022
A transgenic mouse embryonic stem cell line for puromycin selection of V0V interneurons from heterogenous induced cultures.
Stem cell research & therapy
Pardieck J,Harb M,Sakiyama-Elbert SE
450-03 was used in Cell Culture to generate a transgenic mESC line to enrich V0V INs from induced cultures to allow for a scalable, enriched population for future in vitro and in vivo studies.
Mon Mar 28 00:00:00 UTC 2022
Young CSF restores oligodendrogenesis and memory in aged mice via Fgf17.
Nature
Iram T,Kern F,Kaur A,Myneni S,Morningstar AR,Shin H,Garcia MA,Yerra L,Palovics R,Yang AC,Hahn O,Lu N,Shuken SR,Haney MS,Lehallier B,Iyer M,Luo J,Zetterberg H,Keller A,Zuchero JB,Wyss-Coray T
450-03 was used in Cell Culture to demonstrate the rejuvenating power of young CSF and identify Fgf17 as a key target to restore oligodendrocyte function in the ageing brain.
Sun May 01 00:00:00 UTC 2022
S1-END-seq reveals DNA secondary structures in human cells.
Molecular cell
Matos-Rodrigues G,van Wietmarschen N,Wu W,Tripathi V,Koussa NC,Pavani R,Nathan WJ,Callen E,Belinky F,Mohammed A,Napierala M,Usdin K,Ansari AZ,Mirkin SM,Nussenzweig A
450-03 was used in Cell Culture to identify dynamic DNA secondary structures in vivo that contribute to elevated genome instability.
Thu Oct 06 00:00:00 UTC 2022
The Extracellular Matrix Proteins Tenascin-C and Tenascin-R Retard Oligodendrocyte Precursor Maturation and Myelin Regeneration in a Cuprizone-Induced Long-Term Demyelination Animal Model.
Cells
Bauch J,Faissner A
450-03 was used in Cell Culture to study how tenascins interfere with remyelination in vivo.
Sat May 28 00:00:00 UTC 2022
Characterization of Neuronal Lysosome Interactome with Proximity Labeling Proteomics.
Journal of visualized experiments : JoVE
Frankenfield A,Ni J,Hao L
450-03 was used in Cell Culture to demonstrate a detailed protocol for endogenous (knock-in) lysosome proximity labeling proteomic method in human induced pluripotent stem cell (hiPSC)-derived neurons.
Thu Jun 23 00:00:00 UTC 2022
Alzheimer's disease like neuropathology in Down syndrome cortical organoids.
Frontiers in cellular neuroscience
Zhao HH,Haddad GG
450-03 was used in Cell Culture to use induced pluripotent stem cell (iPSC) and iPSC-derived 3D cortical organoids to model Alzheimer's disease in Down syndrome and explore the earliest cellular and molecular changes during DS fetal brain development.
Tue Jan 03 00:00:00 UTC 2023
Pharmacological Perturbation of Mechanical Contractility Enables Robust Transdifferentiation of Human Fibroblasts into Neurons.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
He ZQ,Li YH,Feng GH,Yuan XW,Lu ZB,Dai M,Hu YP,Zhang Y,Zhou Q,Li W
450-03 was used in Cell Culture to report a robust method to convert fibroblasts to neurons through targeting the cytoskeleton followed by exposure to lineage-specification surroundings.
Sun May 01 00:00:00 UTC 2022
CNS myelination requires VAMP2/3-mediated membrane expansion in oligodendrocytes.
Nature communications
Lam M,Takeo K,Almeida RG,Cooper MH,Wu K,Iyer M,Kantarci H,Zuchero JB
450-03 was used in Cell Culture to study how CNS myelin membrane addition requires exocytosis mediated by the vesicular SNARE proteins VAMP2/3.
Fri Sep 23 00:00:00 UTC 2022
CNS myelination requires VAMP2/3-mediated membrane expansion in oligodendrocytes.
Nature communications
Lam M,Takeo K,Almeida RG,Cooper MH,Wu K,Iyer M,Kantarci H,Zuchero JB
450-03 was used in Cell Culture to study how CNS myelin membrane addition requires exocytosis mediated by the vesicular SNARE proteins VAMP2/3.
Fri Sep 23 00:00:00 UTC 2022
Brain-derived autophagosome profiling reveals the engulfment of nucleoid-enriched mitochondrial fragments by basal autophagy in neurons.
Neuron
Goldsmith J,Ordureau A,Harper JW,Holzbaur ELF
450-03 was used in Cell Culture to propose that localized Drp1-dependent fission of nucleoid-enriched fragments in proximity to the sites of autophagosome biogenesis enhances their capture.
Wed Mar 16 00:00:00 UTC 2022
Evidence that direct inhibition of transcription factor binding is the prevailing mode of gene and repeat repression by DNA methylation.
Nature genetics
Kaluscha S,Domcke S,Wirbelauer C,Stadler MB,Durdu S,Burger L,Schübeler D
450-03 was used in Cell Culture to reveal methylation-sensitive TFs in vivo and argues that direct inhibition, rather than indirect repression by the tested MBD proteins, is the prevailing mechanism of methylation-mediated repression at regulatory regions and repeats.
Thu Dec 01 00:00:00 UTC 2022
Veteran-derived cerebral organoids display multifaceted pathological defects in studies on Gulf War Illness.
Frontiers in cellular neuroscience
Yates PL,Case K,Sun X,Sullivan K,Baas PW,Qiang L
450-03 was used in Cell Culture to suggest that veteran-derived human cerebral organoids not only can be used as an innovative human model to uncover the cellular responses to GW toxicants but can also serve as a platform for developing personalized medicine approaches for the veterans.
Wed Jan 11 00:00:00 UTC 2023
Culturing and Neuronal Differentiation of Human Dental Pulp Stem Cells.
Current protocols
Goorha S,Victor AK,Reiter LT
450-03 was used in In vitro experiments to provide our protocol for the differentiation of early passage DPSC cultures into neurons for molecular and cellular studies.
Tue Nov 01 00:00:00 UTC 2022
Tissue- and cell-type-specific molecular and functional signatures of 16p11.2 reciprocal genomic disorder across mouse brain and human neuronal models.
American journal of human genetics
Tai DJC,Razaz P,Erdin S,Gao D,Wang J,Nuttle X,de Esch CE,Collins RL,Currall BB,O'Keefe K,Burt ND,Yadav R,Wang L,Mohajeri K,Aneichyk T,Ragavendran A,Stortchevoi A,Morini E,Ma W,Lucente D,Hastie A,Kelleher RJ,Perlis RH,Talkowski ME,Gusella JF
450-03 was used in In vitro experiments to indicate the genomic disorder involves disruption of multiple contributing biological processes and that this disruption has relative impacts that are context specific.
Thu Oct 06 00:00:00 UTC 2022
Neurotoxic reactive astrocytes induce cell death via saturated lipids.
Nature
Guttenplan KA,Weigel MK,Prakash P,Wijewardhane PR,Hasel P,Rufen-Blanchette U,Münch AE,Blum JA,Fine J,Neal MC,Bruce KD,Gitler AD,Chopra G,Liddelow SA,Barres BA
Mon Nov 01 00:00:00 UTC 2021
Genome-wide programmable transcriptional memory by CRISPR-based epigenome editing.
Cell
Nuñez JK,Chen J,Pommier GC,Cogan JZ,Replogle JM,Adriaens C,Ramadoss GN,Shi Q,Hung KL,Samelson AJ,Pogson AN,Kim JYS,Chung A,Leonetti MD,Chang HY,Kampmann M,Bernstein BE,Hovestadt V,Gilbert LA,Weissman JS
Thu Apr 29 00:00:00 UTC 2021
ELAVL4, splicing, and glutamatergic dysfunction precede neuron loss in MAPT mutation cerebral organoids.
Cell
Bowles KR,Silva MC,Whitney K,Bertucci T,Berlind JE,Lai JD,Garza JC,Boles NC,Mahali S,Strang KH,Marsh JA,Chen C,Pugh DA,Liu Y,Gordon RE,Goderie SK,Chowdhury R,Lotz S,Lane K,Crary JF,Haggarty SJ,Karch CM,Ichida JK,Goate AM,Temple S
Thu Aug 19 00:00:00 UTC 2021
H3.3-K27M drives neural stem cell-specific gliomagenesis in a human iPSC-derived model.
Cancer cell
Haag D,Mack N,Benites Goncalves da Silva P,Statz B,Clark J,Tanabe K,Sharma T,Jäger N,Jones DTW,Kawauchi D,Wernig M,Pfister SM
Mon Mar 08 00:00:00 UTC 2021
Long-term maturation of human cortical organoids matches key early postnatal transitions.
Nature neuroscience
Gordon A,Yoon SJ,Tran SS,Makinson CD,Park JY,Andersen J,Valencia AM,Horvath S,Xiao X,Huguenard JR,Pașca SP,Geschwind DH
450-03 was used in Cell Culture to suggest that important components of an intrinsic in vivo developmental program persist in vitro.
Mon Mar 01 00:00:00 UTC 2021
Deconstructing Stepwise Fate Conversion of Human Fibroblasts to Neurons by MicroRNAs.
Cell stem cell
Cates K,McCoy MJ,Kwon JS,Liu Y,Abernathy DG,Zhang B,Liu S,Gontarz P,Kim WK,Chen S,Kong W,Ho JN,Burbach KF,Gabel HW,Morris SA,Yoo AS
450-03 was used in Sample Preparation to study the activity of microRNAs miR-9/9∗ and miR-124 (miR-9/9∗-124) as reprogramming agents which rchestrate direct conversion of human fibroblasts into motor neurons.
Thu Jan 07 00:00:00 UTC 2021
Deconstructing Stepwise Fate Conversion of Human Fibroblasts to Neurons by MicroRNAs.
Cell stem cell
Cates K,McCoy MJ,Kwon JS,Liu Y,Abernathy DG,Zhang B,Liu S,Gontarz P,Kim WK,Chen S,Kong W,Ho JN,Burbach KF,Gabel HW,Morris SA,Yoo AS
450-03 was used in Sample Preparation to study the activity of microRNAs miR-9/9∗ and miR-124 (miR-9/9∗-124) as reprogramming agents which rchestrate direct conversion of human fibroblasts into motor neurons.
Thu Jan 07 00:00:00 UTC 2021
Temporal proteomics during neurogenesis reveals large-scale proteome and organelle remodeling via selective autophagy.
Molecular cell
Ordureau A,Kraus F,Zhang J,An H,Park S,Ahfeldt T,Paulo JA,Harper JW
Thu Dec 16 00:00:00 UTC 2021
Induced pluripotent stem cell-derived motor neurons of CMT type 2 patients reveal progressive mitochondrial dysfunction.
Brain : a journal of neurology
Van Lent J,Verstraelen P,Asselbergh B,Adriaenssens E,Mateiu L,Verbist C,De Winter V,Eggermont K,Van Den Bosch L,De Vos WH,Timmerman V
450-03 was used in Cell Culture to reveal shared cellular phenotypes across different CMT2 subtypes and suggests that targeting such common pathomechanisms could allow the development of a uniform treatment for CMT2.
Sat Sep 04 00:00:00 UTC 2021
LAG3 is not expressed in human and murine neurons and does not modulate α-synucleinopathies.
EMBO molecular medicine
Emmenegger M,De Cecco E,Hruska-Plochan M,Eninger T,Schneider MM,Barth M,Tantardini E,de Rossi P,Bacioglu M,Langston RG,Kaganovich A,Bengoa-Vergniory N,Gonzalez-Guerra A,Avar M,Heinzer D,Reimann R,Häsler LM,Herling TW,Matharu NS,Landeck N,Luk K,Melki R,Kahle PJ,Hornemann S,Knowles TPJ,Cookson MR,Polymenidou M,Jucker M,Aguzzi A
Tue Sep 07 00:00:00 UTC 2021
Pharmacological reversal of synaptic and network pathology in human MECP2-KO neurons and cortical organoids.
EMBO molecular medicine
Trujillo CA,Adams JW,Negraes PD,Carromeu C,Tejwani L,Acab A,Tsuda B,Thomas CA,Sodhi N,Fichter KM,Romero S,Zanella F,Sejnowski TJ,Ulrich H,Muotri AR
Mon Jan 11 00:00:00 UTC 2021
BNIP3L-mediated mitophagy is required for mitochondrial remodeling during the differentiation of optic nerve oligodendrocytes.
Autophagy
Yazdankhah M,Ghosh S,Shang P,Stepicheva N,Hose S,Liu H,Chamling X,Tian S,Sullivan MLG,Calderon MJ,Fitting CS,Weiss J,Jayagopal A,Handa JT,Sahel JA,Zigler JS,Kinchington PR,Zack DJ,Sinha D
450-03 was used in Sample Preparation (FC/FACS) to investigate mechanisms of oligodendrocyte progenitor cell differentiation to identify treatment options.
Fri Oct 01 00:00:00 UTC 2021
Increased LRRK2 kinase activity alters neuronal autophagy by disrupting the axonal transport of autophagosomes.
Current biology : CB
Boecker CA,Goldsmith J,Dou D,Cajka GG,Holzbaur ELF
Mon May 24 00:00:00 UTC 2021
Human cell transformation by combined lineage conversion and oncogene expression.
Oncogene
Sahu B,Pihlajamaa P,Zhang K,Palin K,Ahonen S,Cervera A,Ristimäki A,Aaltonen LA,Hautaniemi S,Taipale J
Wed Sep 01 00:00:00 UTC 2021
Premigratory neural crest stem cells generate enteric neurons populating the mouse colon and regulating peristalsis in tissue-engineered intestine.
Stem cells translational medicine
Yuan H,Hu H,Chen R,Mu W,Wang L,Li Y,Chen Y,Ding X,Xi Y,Mao S,Jiang M,Chen J,He Y,Wang L,Dong Y,Tou J,Chen W
Tue Jun 01 00:00:00 UTC 2021
The thrombin receptor links brain derived neurotrophic factor to neuron cholesterol production, resiliency and repair after spinal cord injury.
Neurobiology of disease
Triplet EM,Kim HN,Yoon H,Radulovic M,Kleppe L,Simon WL,Choi CI,Walsh PJ,Dutton JR,Scarisbrick IA
450-03 was used in In vitro experiments to identify the thrombin receptor (Protease Activated Receptor 1 (PAR1)) as an integral facet of this machine with roles in regulating neurite growth through a growth factor- and cholesterol-dependent mechanism.
Sat May 01 00:00:00 UTC 2021
Extracellular Vesicles in Serum and Central Nervous System Tissues Contain microRNA Signatures in Sporadic Amyotrophic Lateral Sclerosis.
Frontiers in molecular neuroscience
Lo TW,Figueroa-Romero C,Hur J,Pacut C,Stoll E,Spring C,Lewis R,Nair A,Goutman SA,Sakowski SA,Nagrath S,Feldman EL
450-03 was used in In vitro experiments to study the role of circulating extracellular vesicles in the pathogenesis of amyotrophic lateral sclerosis.
Wed Nov 17 00:00:00 UTC 2021
Allele-Specific Gene Editing Rescues Pathology in a Human Model of Charcot-Marie-Tooth Disease Type 2E.
Frontiers in cell and developmental biology
Feliciano CM,Wu K,Watry HL,Marley CBE,Ramadoss GN,Ghanim HY,Liu AZ,Zholudeva LV,McDevitt TC,Saporta MA,Conklin BR,Judge LM
450-03 was used in Cell Culture to study how allele-specific CRISPR gene editing in a human model of axonal Charcot-Marie-Tooth (CMT) disease rescues pathology caused by a dominant missense mutation in the neurofilament light chain gene (NEFL, CMT type 2E).
Thu Sep 07 00:00:00 UTC 2023
Human Induced Pluripotent Stem Cell Derived Sensory Neurons are Sensitive to the Neurotoxic Effects of Paclitaxel.
Clinical and translational science
Xiong C,Chua KC,Stage TB,Priotti J,Kim J,Altman-Merino A,Chan D,Saraf K,Canato Ferracini A,Fattahi F,Kroetz DL
Mon Mar 01 00:00:00 UTC 2021
Induction of Ventral Spinal V0 Interneurons from Mouse Embryonic Stem Cells.
Stem cells and development
Pardieck J,Harb M,Sakiyama-Elbert S
450-03 was used in Cell Culture to investigate V0 interneuron isolation to improve understanding of spinal central pattern generators.
Sun Aug 15 00:00:00 UTC 2021
An iPSC model of hereditary sensory neuropathy-1 reveals L-serine-responsive deficits in neuronal ganglioside composition and axoglial interactions.
Cell reports. Medicine
Clark AJ,Kugathasan U,Baskozos G,Priestman DA,Fugger N,Lone MA,Othman A,Chu KH,Blesneac I,Wilson ER,Laurà M,Kalmar B,Greensmith L,Hornemann T,Platt FM,Reilly MM,Bennett DL
Tue Jul 20 00:00:00 UTC 2021
One-step Reprogramming of Human Fibroblasts into Oligodendrocyte-like Cells by SOX10, OLIG2, and NKX6.2.
Stem cell reports
Chanoumidou K,Hernández-Rodríguez B,Windener F,Thomas C,Stehling M,Mozafari S,Albrecht S,Ottoboni L,Antel J,Kim KP,Velychko S,Cui QL,Xu YKT,Martino G,Winkler J,Schöler HR,Baron-Van Evercooren A,Boespflug-Tanguy O,Vaquerizas JM,Ehrlich M,Kuhlmann T
Tue Apr 13 00:00:00 UTC 2021
SOX10 ablation severely impairs the generation of postmigratory neural crest from human pluripotent stem cells.
Cell death & disease
Lai X,Liu J,Zou Z,Wang Y,Wang Y,Liu X,Huang W,Ma Y,Chen Q,Li F,Wu G,Li W,Wang W,Yuan Y,Jiang B
Fri Aug 27 00:00:00 UTC 2021
High-content image-based analysis and proteomic profiling identifies Tau phosphorylation inhibitors in a human iPSC-derived glutamatergic neuronal model of tauopathy.
Scientific reports
Cheng C,Reis SA,Adams ET,Fass DM,Angus SP,Stuhlmiller TJ,Richardson J,Olafson H,Wang ET,Patnaik D,Beauchamp RL,Feldman DA,Silva MC,Sur M,Johnson GL,Ramesh V,Miller BL,Temple S,Kosik KS,Dickerson BC,Haggarty SJ
Mon Aug 23 00:00:00 UTC 2021
Heterogeneity of neurons reprogrammed from spinal cord astrocytes by the proneural factors Ascl1 and Neurogenin2.
Cell reports
Kempf J,Knelles K,Hersbach BA,Petrik D,Riedemann T,Bednarova V,Janjic A,Simon-Ebert T,Enard W,Smialowski P,Götz M,Masserdotti G
Tue Jul 20 00:00:00 UTC 2021
Select neurotrophins promote oligodendrocyte progenitor cell process outgrowth in the presence of chondroitin sulfate proteoglycans.
Journal of neuroscience research
Siebert JR,Osterhout DJ
Thu Apr 01 00:00:00 UTC 2021
Identification of Novel Positive Allosteric Modulators of Neurotrophin Receptors for the Treatment of Cognitive Dysfunction.
Cells
Dahlström M,Madjid N,Nordvall G,Halldin MM,Vazquez-Juarez E,Lindskog M,Sandin J,Winblad B,Eriksdotter M,Forsell P
450-03 was used in In vitro experiments to identify small molecules with stimulatory effects on the signaling of two neurotrophins, the nerve growth factor (NGF) and the brain derived neurotrophic factor (BDNF).
Fri Jul 23 00:00:00 UTC 2021
Genome-wide CRISPR screen identifies protein pathways modulating tau protein levels in neurons.
Communications biology
Sanchez CG,Acker CM,Gray A,Varadarajan M,Song C,Cochran NR,Paula S,Lindeman A,An S,McAllister G,Alford J,Reece-Hoyes J,Russ C,Craig L,Capre K,Doherty C,Hoffman GR,Luchansky SJ,Polydoro M,Dolmetsch R,Elwood F
Mon Jun 14 00:00:00 UTC 2021
MeCP2 controls neural stem cell fate specification through miR-199a-mediated inhibition of BMP-Smad signaling.
Cell reports
Nakashima H,Tsujimura K,Irie K,Imamura T,Trujillo CA,Ishizu M,Uesaka M,Pan M,Noguchi H,Okada K,Aoyagi K,Andoh-Noda T,Okano H,Muotri AR,Nakashima K
Tue May 18 00:00:00 UTC 2021
BASP1 labels neural stem cells in the neurogenic niches of mammalian brain.
Scientific reports
Manganas LN,Durá I,Osenberg S,Semerci F,Tosun M,Mishra R,Parkitny L,Encinas JM,Maletic-Savatic M
Wed Mar 10 00:00:00 UTC 2021
Overcoming the inhibitory microenvironment surrounding oligodendrocyte progenitor cells following experimental demyelination.
Nature communications
Saraswat D,Shayya HJ,Polanco JJ,Tripathi A,Welliver RR,Pol SU,Seidman RA,Broome JE,O'Bara MA,van Kuppervelt TH,Phillips JJ,Dutta R,Sim FJ
Fri Mar 26 00:00:00 UTC 2021
Deep learning-based predictive identification of neural stem cell differentiation.
Nature communications
Zhu Y,Huang R,Wu Z,Song S,Cheng L,Zhu R
450-03 was used in Cell Culture to showcase effectiveness of deep learning-based platform in precisely, accurately and robustly detecting NSC differentiation.
Mon May 10 00:00:00 UTC 2021
Deep learning-based predictive identification of neural stem cell differentiation.
Nature communications
Zhu Y,Huang R,Wu Z,Song S,Cheng L,Zhu R
450-03 was used in Cell Culture to showcase effectiveness of deep learning-based platform in precisely, accurately and robustly detecting NSC differentiation.
Mon May 10 00:00:00 UTC 2021
SETBP1 accumulation induces P53 inhibition and genotoxic stress in neural progenitors underlying neurodegeneration in Schinzel-Giedion syndrome.
Nature communications
Banfi F,Rubio A,Zaghi M,Massimino L,Fagnocchi G,Bellini E,Luoni M,Cancellieri C,Bagliani A,Di Resta C,Maffezzini C,Ianielli A,Ferrari M,Piazza R,Mologni L,Broccoli V,Sessa A
Wed Jun 30 00:00:00 UTC 2021
5-HT3 Signaling Alters Development of Sacral Neural Crest Derivatives That Innervate the Lower Urinary Tract.
International journal of molecular sciences
Ritter KE,Buehler DP,Asher SB,Deal KK,Zhao S,Guo Y,Southard-Smith EM
450-03 was used in Cell Culture to study the development of pelvic ganglia neuronal diversity.
Fri Jun 25 00:00:00 UTC 2021
Development of a specific live-cell assay for native autophagic flux.
The Journal of biological chemistry
Safren N,Tank EM,Malik AM,Chua JP,Santoro N,Barmada SJ
Wed Sep 01 00:00:00 UTC 2021
STING regulates peripheral nerve regeneration and colony stimulating factor 1 receptor (CSF1R) processing in microglia.
iScience
Morozzi G,Rothen J,Toussaint G,De Lange K,Westritschnig K,Doelemeyer A,Ueberschlag VP,Kahle P,Lambert C,Obrecht M,Beckmann N,Ritter V,Panesar M,Stauffer D,Garnier I,Mueller M,Guerini D,Keller CG,Knehr J,Roma G,Bidinosti M,Brachat S,Morvan F,Fornaro M
450-03 was used in In vitro experiments to investigate relationship between peripheral nerve injury and inflammation.
Fri Dec 17 00:00:00 UTC 2021
Neuronal defects in a human cellular model of 22q11.2 deletion syndrome.
Nature medicine
Khan TA,Revah O,Gordon A,Yoon SJ,Krawisz AK,Goold C,Sun Y,Kim CH,Tian Y,Li MY,Schaepe JM,Ikeda K,Amin ND,Sakai N,Yazawa M,Kushan L,Nishino S,Porteus MH,Rapoport JL,Bernstein JA,O'Hara R,Bearden CE,Hallmayer JF,Huguenard JR,Geschwind DH,Dolmetsch RE,Paşca SP
450-03 was used in Cell Culture to illustrate how stem cell derived models can be used to uncover and rescue cellular phenotypes associated with genetic forms of neuropsychiatric disease.
Tue Dec 01 00:00:00 UTC 2020
Origins and Proliferative States of Human Oligodendrocyte Precursor Cells.
Cell
Huang W,Bhaduri A,Velmeshev D,Wang S,Wang L,Rottkamp CA,Alvarez-Buylla A,Rowitch DH,Kriegstein AR
450-03 was used in Cell Culture to indicate properties of OPC derivation, proliferation, and dispersion important for human white matter expansion and myelination.
Thu Aug 06 00:00:00 UTC 2020
Functionally Distinct Neuronal Ensembles within the Memory Engram.
Cell
Sun X,Bernstein MJ,Meng M,Rao S,Sørensen AT,Yao L,Zhang X,Anikeeva PO,Lin Y
Thu Apr 16 00:00:00 UTC 2020
SARS-CoV-2 infects human neural progenitor cells and brain organoids.
Cell research
Zhang BZ,Chu H,Han S,Shuai H,Deng J,Hu YF,Gong HR,Lee AC,Zou Z,Yau T,Wu W,Hung IF,Chan JF,Yuen KY,Huang JD
Thu Oct 01 00:00:00 UTC 2020
Suppression of MEHMO Syndrome Mutation in eIF2 by Small Molecule ISRIB.
Molecular cell
Young-Baird SK,Lourenço MB,Elder MK,Klann E,Liebau S,Dever TE
450-03 was used in In vitro experiments to show that a mutation at the C terminus of eIF2γ impairs CDC123 promotion of eIF2 complex formation and decreases the level of eIF2-GTP-Met-tRNAiMet ternary complexes, using patient-derived induced pluripotent stem cells.
Thu Feb 20 00:00:00 UTC 2020
Loss of TAX1BP1-Directed Autophagy Results in Protein Aggregate Accumulation in the Brain.
Molecular cell
Sarraf SA,Shah HV,Kanfer G,Pickrell AM,Holtzclaw LA,Ward ME,Youle RJ
450-03 was used in Cell Culture to show that TAX1BP1 mediates clearance of a broad range of cytotoxic proteins indicating therapeutic potential in neurodegenerative diseases.
Thu Dec 03 00:00:00 UTC 2020
Huntington's Disease Pathogenesis Is Modified In Vivo by Alfy/Wdfy3 and Selective Macroautophagy.
Neuron
Fox LM,Kim K,Johnson CW,Chen S,Croce KR,Victor MB,Eenjes E,Bosco JR,Randolph LK,Dragatsis I,Dragich JM,Yoo AS,Yamamoto A
Wed Mar 04 00:00:00 UTC 2020
m6A mRNA Methylation Is Essential for Oligodendrocyte Maturation and CNS Myelination.
Neuron
Xu H,Dzhashiashvili Y,Shah A,Kunjamma RB,Weng YL,Elbaz B,Fei Q,Jones JS,Li YI,Zhuang X,Ming GL,He C,Popko B
Wed Jan 22 00:00:00 UTC 2020
USP7 regulates ALS-associated proteotoxicity and quality control through the NEDD4L-SMAD pathway.
Proceedings of the National Academy of Sciences of the United States of America
Zhang T,Periz G,Lu YN,Wang J
Tue Nov 10 00:00:00 UTC 2020
The formation of paranodal spirals at the ends of CNS myelin sheaths requires the planar polarity protein Vangl2.
Glia
Jarjour AA,Velichkova AN,Boyd A,Lord KM,Torsney C,Henderson DJ,Ffrench-Constant C
Tue Sep 01 00:00:00 UTC 2020
Generation and optimization of highly pure motor neurons from human induced pluripotent stem cells via lentiviral delivery of transcription factors.
American journal of physiology. Cell physiology
Sepehrimanesh M,Ding B
Thu Oct 01 00:00:00 UTC 2020
Multicellular 3D Neurovascular Unit Model for Assessing Hypoxia and Neuroinflammation Induced Blood-Brain Barrier Dysfunction.
Scientific reports
Nzou G,Wicks RT,VanOstrand NR,Mekky GA,Seale SA,El-Taibany A,Wicks EE,Nechtman CM,Marrotte EJ,Makani VS,Murphy SV,Seeds MC,Jackson JD,Atala AJ
Wed Jun 17 00:00:00 UTC 2020
Photoactivatable Cre recombinase 3.0 for in vivo mouse applications.
Nature communications
Morikawa K,Furuhashi K,de Sena-Tomas C,Garcia-Garcia AL,Bekdash R,Klein AD,Gallerani N,Yamamoto HE,Park SE,Collins GS,Kawano F,Sato M,Lin CS,Targoff KL,Au E,Salling MC,Yazawa M
Fri May 01 00:00:00 UTC 2020
Protocol for Tri-culture of hiPSC-Derived Neurons, Astrocytes, and Microglia.
STAR protocols
Ryan SK,Jordan-Sciutto KL,Anderson SA
Fri Dec 18 00:00:00 UTC 2020
Neuroinflammation and EIF2 Signaling Persist despite Antiretroviral Treatment in an hiPSC Tri-culture Model of HIV Infection.
Stem cell reports
Ryan SK,Gonzalez MV,Garifallou JP,Bennett FC,Williams KS,Sotuyo NP,Mironets E,Cook K,Hakonarson H,Anderson SA,Jordan-Sciutto KL
Tue Apr 14 00:00:00 UTC 2020
Identification of candidate genetic variants and altered protein expression in neural stem and mature neural cells support altered microtubule function to be an essential component in bipolar disorder.
Translational psychiatry
Truvé K,Parris TZ,Vizlin-Hodzic D,Salmela S,Berger E,Ågren H,Funa K
Mon Nov 09 00:00:00 UTC 2020
Frataxin gene editing rescues Friedreich's ataxia pathology in dorsal root ganglia organoid-derived sensory neurons.
Nature communications
Mazzara PG,Muggeo S,Luoni M,Massimino L,Zaghi M,Valverde PT,Brusco S,Marzi MJ,Palma C,Colasante G,Iannielli A,Paulis M,Cordiglieri C,Giannelli SG,Podini P,Gellera C,Taroni F,Nicassio F,Rasponi M,Broccoli V
Fri Aug 21 00:00:00 UTC 2020
Transplantation of hPSC-derived pericyte-like cells promotes functional recovery in ischemic stroke mice.
Nature communications
Sun J,Huang Y,Gong J,Wang J,Fan Y,Cai J,Wang Y,Qiu Y,Wei Y,Xiong C,Chen J,Wang B,Ma Y,Huang L,Chen X,Zheng S,Huang W,Ke Q,Wang T,Li X,Zhang W,Xiang AP,Li W
Thu Oct 15 00:00:00 UTC 2020
Cofilin dysregulation alters actin turnover in frataxin-deficient neurons.
Scientific reports
Muñoz-Lasso DC,Mollá B,Calap-Quintana P,García-Giménez JL,Pallardo FV,Palau F,Gonzalez-Cabo P
Mon Mar 23 00:00:00 UTC 2020
Master Regulators and Cofactors of Human Neuronal Cell Fate Specification Identified by CRISPR Gene Activation Screens.
Cell reports
Black JB,McCutcheon SR,Dube S,Barrera A,Klann TS,Rice GA,Adkar SS,Soderling SH,Reddy TE,Gersbach CA
Tue Dec 01 00:00:00 UTC 2020
Generation of oligodendrocytes and establishment of an all-human myelinating platform from human pluripotent stem cells.
Nature protocols
García-León JA,García-Díaz B,Eggermont K,Cáceres-Palomo L,Neyrinck K,Madeiro da Costa R,Dávila JC,Baron-Van Evercooren A,Gutiérrez A,Verfaillie CM
Sun Nov 01 00:00:00 UTC 2020
Transcriptional repression of PTEN in neural cells using CRISPR/dCas9 epigenetic editing.
Scientific reports
Moses C,Hodgetts SI,Nugent F,Ben-Ary G,Park KK,Blancafort P,Harvey AR
Thu Jul 09 00:00:00 UTC 2020
Varicella-zoster virus VLT-ORF63 fusion transcript induces broad viral gene expression during reactivation from neuronal latency.
Nature communications
Ouwendijk WJD,Depledge DP,Rajbhandari L,Lenac Rovis T,Jonjic S,Breuer J,Venkatesan A,Verjans GMGM,Sadaoka T
Thu Dec 10 00:00:00 UTC 2020
Sequentially induced motor neurons from human fibroblasts facilitate locomotor recovery in a rodent spinal cord injury model.
eLife
Lee H,Lee HY,Lee BE,Gerovska D,Park SY,Zaehres H,Araúzo-Bravo MJ,Kim JI,Ha Y,Schöler HR,Kim JB
450-03 was used in Cell Culture to investigate the use of induced motor neurons to treat spinal cord injury.
Tue Jun 23 00:00:00 UTC 2020
Identification of evolutionarily conserved gene networks mediating neurodegenerative dementia.
Nature medicine
Swarup V,Hinz FI,Rexach JE,Noguchi KI,Toyoshiba H,Oda A,Hirai K,Sarkar A,Seyfried NT,Cheng C,Haggarty SJ,Grossman M,Van Deerlin VM,Trojanowski JQ,Lah JJ,Levey AI,Kondou S,Geschwind DH
450-03 was used in Cell Culture to identify small molecules that can normalize the disease-associated modules.
Tue Jan 01 00:00:00 UTC 2019
Neuromodulator Signaling Bidirectionally Controls Vesicle Numbers in Human Synapses.
Cell
Patzke C,Brockmann MM,Dai J,Gan KJ,Grauel MK,Fenske P,Liu Y,Acuna C,Rosenmund C,Südhof TC
Thu Oct 03 00:00:00 UTC 2019
RNA Granules Hitchhike on Lysosomes for Long-Distance Transport, Using Annexin A11 as a Molecular Tether.
Cell
Liao YC,Fernandopulle MS,Wang G,Choi H,Hao L,Drerup CM,Patel R,Qamar S,Nixon-Abell J,Shen Y,Meadows W,Vendruscolo M,Knowles TPJ,Nelson M,Czekalska MA,Musteikyte G,Gachechiladze MA,Stephens CA,Pasolli HA,Forrest LR,St George-Hyslop P,Lippincott-Schwartz J,Ward ME
Thu Sep 19 00:00:00 UTC 2019
Methotrexate Chemotherapy Induces Persistent Tri-glial Dysregulation that Underlies Chemotherapy-Related Cognitive Impairment.
Cell
Gibson EM,Nagaraja S,Ocampo A,Tam LT,Wood LS,Pallegar PN,Greene JJ,Geraghty AC,Goldstein AK,Ni L,Woo PJ,Barres BA,Liddelow S,Vogel H,Monje M
Thu Jan 10 00:00:00 UTC 2019
Reliability of human cortical organoid generation.
Nature methods
Yoon SJ,Elahi LS,Pașca AM,Marton RM,Gordon A,Revah O,Miura Y,Walczak EM,Holdgate GM,Fan HC,Huguenard JR,Geschwind DH,Pașca SP
450-03 was used in Cell Culture to find that spheroid generation was highly reliable and consistent.
Tue Jan 01 00:00:00 UTC 2019
Increased synapse elimination by microglia in schizophrenia patient-derived models of synaptic pruning.
Nature neuroscience
Sellgren CM,Gracias J,Watmuff B,Biag JD,Thanos JM,Whittredge PB,Fu T,Worringer K,Brown HE,Wang J,Kaykas A,Karmacharya R,Goold CP,Sheridan SD,Perlis RH
450-03 was used in Cell Culture to demonstrate increased synapse elimination in patient-derived neural cultures and isolated synaptosomes.
Fri Mar 01 00:00:00 UTC 2019
Oligodendrocyte Death in Pelizaeus-Merzbacher Disease Is Rescued by Iron Chelation.
Cell stem cell
Nobuta H,Yang N,Ng YH,Marro SG,Sabeur K,Chavali M,Stockley JH,Killilea DW,Walter PB,Zhao C,Huie P,Goldman SA,Kriegstein AR,Franklin RJM,Rowitch DH,Wernig M
450-03 was used in Cell Culture to demonstrate that patient-derived oligodendrocytes can develop to the pre-myelinating stage, but subsequently undergo cell death using induced pluripotent stem cell (iPSC) and gene-correction.
Thu Oct 03 00:00:00 UTC 2019
Loss of Adaptive Myelination Contributes to Methotrexate Chemotherapy-Related Cognitive Impairment.
Neuron
Geraghty AC,Gibson EM,Ghanem RA,Greene JJ,Ocampo A,Goldstein AK,Ni L,Yang T,Marton RM,Paşca SP,Greenberg ME,Longo FM,Monje M
Wed Jul 17 00:00:00 UTC 2019
Loss of Adaptive Myelination Contributes to Methotrexate Chemotherapy-Related Cognitive Impairment.
Neuron
Geraghty AC,Gibson EM,Ghanem RA,Greene JJ,Ocampo A,Goldstein AK,Ni L,Yang T,Marton RM,Paşca SP,Greenberg ME,Longo FM,Monje M
Wed Jul 17 00:00:00 UTC 2019
Defining the Functional Role of NaV1.7 in Human Nociception.
Neuron
McDermott LA,Weir GA,Themistocleous AC,Segerdahl AR,Blesneac I,Baskozos G,Clark AJ,Millar V,Peck LJ,Ebner D,Tracey I,Serra J,Bennett DL
Wed Mar 06 00:00:00 UTC 2019
Pathogenic Tau Impairs Axon Initial Segment Plasticity and Excitability Homeostasis.
Neuron
Sohn PD,Huang CT,Yan R,Fan L,Tracy TE,Camargo CM,Montgomery KM,Arhar T,Mok SA,Freilich R,Baik J,He M,Gong S,Roberson ED,Karch CM,Gestwicki JE,Xu K,Kosik KS,Gan L
Wed Nov 06 00:00:00 UTC 2019
Nucleoporin Seh1 Interacts with Olig2/Brd7 to Promote Oligodendrocyte Differentiation and Myelination.
Neuron
Liu Z,Yan M,Liang Y,Liu M,Zhang K,Shao D,Jiang R,Li L,Wang C,Nussenzveig DR,Zhang K,Chen S,Zhong C,Mo W,Fontoura BMA,Zhang L
Wed May 08 00:00:00 UTC 2019
CRISPR Interference-Based Platform for Multimodal Genetic Screens in Human iPSC-Derived Neurons.
Neuron
Tian R,Gachechiladze MA,Ludwig CH,Laurie MT,Hong JY,Nathaniel D,Prabhu AV,Fernandopulle MS,Patel R,Abshari M,Ward ME,Kampmann M
Wed Oct 23 00:00:00 UTC 2019
Pioneer Factor NeuroD1 Rearranges Transcriptional and Epigenetic Profiles to Execute Microglia-Neuron Conversion.
Neuron
Matsuda T,Irie T,Katsurabayashi S,Hayashi Y,Nagai T,Hamazaki N,Adefuin AMD,Miura F,Ito T,Kimura H,Shirahige K,Takeda T,Iwasaki K,Imamura T,Nakashima K
Wed Feb 06 00:00:00 UTC 2019
A new patient-derived iPSC model for dystroglycanopathies validates a compound that increases glycosylation of α-dystroglycan.
EMBO reports
Kim J,Lana B,Torelli S,Ryan D,Catapano F,Ala P,Luft C,Stevens E,Konstantinidis E,Louzada S,Fu B,Paredes-Redondo A,Chan AE,Yang F,Stemple DL,Liu P,Ketteler R,Selwood DL,Muntoni F,Lin YY
Tue Nov 05 00:00:00 UTC 2019
Astrocyte-to-astrocyte contact and a positive feedback loop of growth factor signaling regulate astrocyte maturation.
Glia
Li J,Khankan RR,Caneda C,Godoy MI,Haney MS,Krawczyk MC,Bassik MC,Sloan SA,Zhang Y
Thu Aug 01 00:00:00 UTC 2019
Perfluorocarbon Labeling of Human Glial-Restricted Progenitors for 19 F Magnetic Resonance Imaging.
Stem cells translational medicine
Richard JP,Hussain U,Gross S,Taga A,Kouser M,Almad A,Campanelli JT,Bulte JWM,Maragakis NJ
Mon Apr 01 00:00:00 UTC 2019
Chromatin Decondensation by FOXP2 Promotes Human Neuron Maturation and Expression of Neurodevelopmental Disease Genes.
Cell reports
Hickey SL,Berto S,Konopka G
450-03 was used in Cell Culture to study the mechanisms by which FOXP2 exerts regulatory control over targets during neuronal maturation.
Tue May 07 00:00:00 UTC 2019
GAIN domain-mediated cleavage is required for activation of G protein-coupled receptor 56 (GPR56) by its natural ligands and a small-molecule agonist.
The Journal of biological chemistry
Zhu B,Luo R,Jin P,Li T,Oak HC,Giera S,Monk KR,Lak P,Shoichet BK,Piao X
Fri Dec 13 00:00:00 UTC 2019
Harnessing neurovascular interaction to guide axon growth.
Scientific reports
Partyka PP,Jin Y,Bouyer J,DaSilva A,Godsey GA,Nagele RG,Fischer I,Galie PA
Mon Feb 18 00:00:00 UTC 2019
Harnessing neurovascular interaction to guide axon growth.
Scientific reports
Partyka PP,Jin Y,Bouyer J,DaSilva A,Godsey GA,Nagele RG,Fischer I,Galie PA
Mon Feb 18 00:00:00 UTC 2019
A cell fitness selection model for neuronal survival during development.
Nature communications
Wang Y,Wu H,Fontanet P,Codeluppi S,Akkuratova N,Petitpré C,Xue-Franzén Y,Niederreither K,Sharma A,Da Silva F,Comai G,Agirman G,Palumberi D,Linnarsson S,Adameyko I,Moqrich A,Schedl A,La Manno G,Hadjab S,Lallemend F
Thu Sep 12 00:00:00 UTC 2019
Assessing the therapeutic potential of Graptopetalum paraguayense on Alzheimer's disease using patient iPSC-derived neurons.
Scientific reports
Wu PC,Fann MJ,Tran TT,Chen SC,Devina T,Cheng IH,Lien CC,Kao LS,Wang SJ,Fuh JL,Tzeng TT,Huang CY,Shiao YJ,Wong YH
Tue Dec 17 00:00:00 UTC 2019
Postinjury Inhibition of miR-181a Promotes Restoration of Hippocampal CA1 Neurons after Transient Forebrain Ischemia in Rats.
eNeuro
Griffiths BB,Ouyang YB,Xu L,Sun X,Giffard RG,Stary CM
450-03 was used in In vitro experiments to evaluate the use of anti-miR-181a for postinjury treatment of CA1 neuron recovery.
Thu Mar 05 00:00:00 UTC 2020
Oligodendrocyte Intrinsic miR-27a Controls Myelination and Remyelination.
Cell reports
Tripathi A,Volsko C,Garcia JP,Agirre E,Allan KC,Tesar PJ,Trapp BD,Castelo-Branco G,Sim FJ,Dutta R
450-03 was used in Cell Culture to provide evidence supporting a critical role for a steady-state level of OL-specific miR-27a in supporting multiple steps in the complex process of OPC maturation and remyelination.
Tue Oct 22 00:00:00 UTC 2019
Oligodendrocyte Intrinsic miR-27a Controls Myelination and Remyelination.
Cell reports
Tripathi A,Volsko C,Garcia JP,Agirre E,Allan KC,Tesar PJ,Trapp BD,Castelo-Branco G,Sim FJ,Dutta R
450-03 was used in Cell Culture to provide evidence supporting a critical role for a steady-state level of OL-specific miR-27a in supporting multiple steps in the complex process of OPC maturation and remyelination.
Tue Oct 22 00:00:00 UTC 2019
Autaptic cultures of human induced neurons as a versatile platform for studying synaptic function and neuronal morphology.
Scientific reports
Fenske P,Grauel MK,Brockmann MM,Dorrn AL,Trimbuch T,Rosenmund C
Wed Mar 20 00:00:00 UTC 2019
Stem Cell Modeling of Neuroferritinopathy Reveals Iron as a Determinant of Senescence and Ferroptosis during Neuronal Aging.
Stem cell reports
Cozzi A,Orellana DI,Santambrogio P,Rubio A,Cancellieri C,Giannelli S,Ripamonti M,Taverna S,Di Lullo G,Rovida E,Ferrari M,Forni GL,Fiorillo C,Broccoli V,Levi S
Tue Nov 12 00:00:00 UTC 2019
Binding of α-synuclein oligomers to Cx32 facilitates protein uptake and transfer in neurons and oligodendrocytes.
Acta neuropathologica
Reyes JF,Sackmann C,Hoffmann A,Svenningsson P,Winkler J,Ingelsson M,Hallbeck M
450-03 was used in Cell Culture to study how the gap junction protein connexin-32 (Cx32) is centrally involved in the preferential uptake of α-syn oligomeric assemblies (oα-syn) in neurons and oligodendrocytes to provide compelling evidence for Cx32 as a novel target for therapeutic intervention in Parkinson's disease (PD) and related α-synucleinopathies.
Mon Jul 01 00:00:00 UTC 2019
Precisely controlling endogenous protein dosage in hPSCs and derivatives to model FOXG1 syndrome.
Nature communications
Zhu W,Zhang B,Li M,Mo F,Mi T,Wu Y,Teng Z,Zhou Q,Li W,Hu B
Mon Feb 25 00:00:00 UTC 2019
V2a interneuron differentiation from mouse and human pluripotent stem cells.
Nature protocols
Butts JC,Iyer N,White N,Thompson R,Sakiyama-Elbert S,McDevitt TC
450-03 was used in Cell Culture to describe how to produce V2a interneurons from mouse and human pluripotent stem cells (PSCs), as well as strategies to characterize and mature the cells for further analysis.
Fri Nov 01 00:00:00 UTC 2019
V2a interneuron differentiation from mouse and human pluripotent stem cells.
Nature protocols
Butts JC,Iyer N,White N,Thompson R,Sakiyama-Elbert S,McDevitt TC
450-03 was used in Cell Culture to describe how to produce V2a interneurons from mouse and human pluripotent stem cells (PSCs), as well as strategies to characterize and mature the cells for further analysis.
Fri Nov 01 00:00:00 UTC 2019
Human evolved regulatory elements modulate genes involved in cortical expansion and neurodevelopmental disease susceptibility.
Nature communications
Won H,Huang J,Opland CK,Hartl CL,Geschwind DH
Mon Jun 03 00:00:00 UTC 2019
A Simple Procedure for Creating Scalable Phenotypic Screening Assays in Human Neurons.
Scientific reports
Sridharan B,Hubbs C,Llamosas N,Kilinc M,Singhera FU,Willems E,Piper DR,Scampavia L,Rumbaugh G,Spicer TP
Fri Jun 21 00:00:00 UTC 2019
Biomimicking Fiber Scaffold as an Effective In Vitro and In Vivo MicroRNA Screening Platform for Directing Tissue Regeneration.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zhang N,Milbreta U,Chin JS,Pinese C,Lin J,Shirahama H,Jiang W,Liu H,Mi R,Hoke A,Wu W,Chew SY
Fri May 03 00:00:00 UTC 2019
The Dynamic Landscape of Open Chromatin during Human Cortical Neurogenesis.
Cell
de la Torre-Ubieta L,Stein JL,Won H,Opland CK,Liang D,Lu D,Geschwind DH
Thu Jan 11 00:00:00 UTC 2018
3D Printed Stem-Cell Derived Neural Progenitors Generate Spinal Cord Scaffolds.
Advanced functional materials
Joung D,Truong V,Neitzke CC,Guo SZ,Walsh PJ,Monat JR,Meng F,Park SH,Dutton JR,Parr AM,McAlpine MC
450-03 was used in Cell Culture to introduce a living platform to model spinal cord cytoarchitecture which could ultimately be used as a therapeutic for chronic spinal cord injury to regenerate axons across the lesion site.
Wed Sep 26 00:00:00 UTC 2018
APOE4 Causes Widespread Molecular and Cellular Alterations Associated with Alzheimer's Disease Phenotypes in Human iPSC-Derived Brain Cell Types.
Neuron
Lin YT,Seo J,Gao F,Feldman HM,Wen HL,Penney J,Cam HP,Gjoneska E,Raja WK,Cheng J,Rueda R,Kritskiy O,Abdurrob F,Peng Z,Milo B,Yu CJ,Elmsaouri S,Dey D,Ko T,Yankner BA,Tsai LH
Wed Jun 27 00:00:00 UTC 2018
GPR56/ADGRG1 regulates development and maintenance of peripheral myelin.
The Journal of experimental medicine
Ackerman SD,Luo R,Poitelon Y,Mogha A,Harty BL,D'Rozario M,Sanchez NE,Lakkaraju AKK,Gamble P,Li J,Qu J,MacEwan MR,Ray WZ,Aguzzi A,Feltri ML,Piao X,Monk KR
Mon Mar 05 00:00:00 UTC 2018
Generation and assembly of human brain region-specific three-dimensional cultures.
Nature protocols
Sloan SA,Andersen J,Pașca AM,Birey F,Pașca SP
450-03 was used in In vitro experiments to propose a method to generate and assemble subdomain-specific forebrain spheroids, also known as brain region-specific organoids, from human pluripotent stem cells (hPSCs).
Sat Sep 01 00:00:00 UTC 2018
MicroRNAs Overcome Cell Fate Barrier by Reducing EZH2-Controlled REST Stability during Neuronal Conversion of Human Adult Fibroblasts.
Developmental cell
Lee SW,Oh YM,Lu YL,Kim WK,Yoo AS
450-03 was used in In vitro experiments to study how miR-9/9∗-124 breaks the cell fate barrier to activate the neuronal program.
Mon Jul 02 00:00:00 UTC 2018
Nfat/calcineurin signaling promotes oligodendrocyte differentiation and myelination by transcription factor network tuning.
Nature communications
Weider M,Starost LJ,Groll K,Küspert M,Sock E,Wedel M,Fröb F,Schmitt C,Baroti T,Hartwig AC,Hillgärtner S,Piefke S,Fadler T,Ehrlich M,Ehlert C,Stehling M,Albrecht S,Jabali A,Schöler HR,Winkler J,Kuhlmann T,Wegner M
Fri Mar 02 00:00:00 UTC 2018
Lgl1 controls NG2 endocytic pathway to regulate oligodendrocyte differentiation and asymmetric cell division and gliomagenesis.
Nature communications
Daynac M,Chouchane M,Collins HY,Murphy NE,Andor N,Niu J,Fancy SPJ,Stallcup WB,Petritsch CK
Tue Aug 21 00:00:00 UTC 2018
Preferential amplification of a human mitochondrial DNA deletion in vitro and in vivo.
Scientific reports
Russell OM,Fruh I,Rai PK,Marcellin D,Doll T,Reeve A,Germain M,Bastien J,Rygiel KA,Cerino R,Sailer AW,Lako M,Taylor RW,Mueller M,Lightowlers RN,Turnbull DM,Helliwell SB
Mon Jan 29 00:00:00 UTC 2018
The N-peptide-binding mode is critical to Munc18-1 function in synaptic exocytosis.
The Journal of biological chemistry
Shen C,Liu Y,Yu H,Gulbranson DR,Kogut I,Bilousova G,Zhang C,Stowell MHB,Shen J
Fri Nov 23 00:00:00 UTC 2018
Oxaliplatin induces pH acidification in dorsal root ganglia neurons.
Scientific reports
Riva B,Dionisi M,Potenzieri A,Chiorazzi A,Cordero-Sanchez C,Rigolio R,Carozzi VA,Lim D,Cavaletti G,Marmiroli P,Distasi C,Genazzani AA
Wed Oct 10 00:00:00 UTC 2018
Laminin heparin-binding peptides bind to several growth factors and enhance diabetic wound healing.
Nature communications
Ishihara J,Ishihara A,Fukunaga K,Sasaki K,White MJV,Briquez PS,Hubbell JA
Mon Jun 04 00:00:00 UTC 2018
Laminin heparin-binding peptides bind to several growth factors and enhance diabetic wound healing.
Nature communications
Ishihara J,Ishihara A,Fukunaga K,Sasaki K,White MJV,Briquez PS,Hubbell JA
Mon Jun 04 00:00:00 UTC 2018
Phenotypic and Functional Characterization of Peripheral Sensory Neurons derived from Human Embryonic Stem Cells.
Scientific reports
Alshawaf AJ,Viventi S,Qiu W,D'Abaco G,Nayagam B,Erlichster M,Chana G,Everall I,Ivanusic J,Skafidas E,Dottori M
Fri Jan 12 00:00:00 UTC 2018
Differential antiviral immunity to Japanese encephalitis virus in developing cortical organoids.
Cell death & disease
Zhang B,He Y,Xu Y,Mo F,Mi T,Shen QS,Li C,Li Y,Liu J,Wu Y,Chen G,Zhu W,Qin C,Hu B,Zhou G
Mon Jun 18 00:00:00 UTC 2018
SiO2 nanoparticles modulate the electrical activity of neuroendocrine cells without exerting genomic effects.
Scientific reports
Distasi C,Ruffinatti FA,Dionisi M,Antoniotti S,Gilardino A,Croci G,Riva B,Bassino E,Alberto G,Castroflorio E,Incarnato D,Morandi E,Martra G,Oliviero S,Munaron L,Lovisolo D
Fri Feb 09 00:00:00 UTC 2018
Human Cortex Spheroid with a Functional Blood Brain Barrier for High-Throughput Neurotoxicity Screening and Disease Modeling.
Scientific reports
Nzou G,Wicks RT,Wicks EE,Seale SA,Sane CH,Chen A,Murphy SV,Jackson JD,Atala AJ
Wed May 09 00:00:00 UTC 2018
Chemical conversion of human lung fibroblasts into neuronal cells.
International journal of molecular medicine
Wan XY,Xu LY,Li B,Sun QH,Ji QL,Huang DD,Zhao L,Xiao YT
450-03 was used in Cell Culture to investigate whether a cocktail of small molecules can convert human lung fibroblast cells into neurons.
Thu Mar 01 00:00:00 UTC 2018
Lysosomal and network alterations in human mucopolysaccharidosis type VII iPSC-derived neurons.
Scientific reports
Bayó-Puxan N,Terrasso AP,Creyssels S,Simão D,Begon-Pescia C,Lavigne M,Salinas S,Bernex F,Bosch A,Kalatzis V,Levade T,Cuervo AM,Lory P,Consiglio A,Brito C,Kremer EJ
Fri Nov 09 00:00:00 UTC 2018
Far Upstream Element-Binding Protein 1 Regulates LSD1 Alternative Splicing to Promote Terminal Differentiation of Neural Progenitors.
Stem cell reports
Hwang I,Cao D,Na Y,Kim DY,Zhang T,Yao J,Oh H,Hu J,Zheng H,Yao Y,Paik J
Tue Apr 10 00:00:00 UTC 2018
Assembly of functionally integrated human forebrain spheroids.
Nature
Birey F,Andersen J,Makinson CD,Islam S,Wei W,Huber N,Fan HC,Metzler KRC,Panagiotakos G,Thom N,O'Rourke NA,Steinmetz LM,Bernstein JA,Hallmayer J,Huguenard JR,Paşca SP
Thu May 04 00:00:00 UTC 2017
Fibrinogen Activates BMP Signaling in Oligodendrocyte Progenitor Cells and Inhibits Remyelination after Vascular Damage.
Neuron
Petersen MA,Ryu JK,Chang KJ,Etxeberria A,Bardehle S,Mendiola AS,Kamau-Devers W,Fancy SPJ,Thor A,Bushong EA,Baeza-Raja B,Syme CA,Wu MD,Rios Coronado PE,Meyer-Franke A,Yahn S,Pous L,Lee JK,Schachtrup C,Lassmann H,Huang EJ,Han MH,Absinta M,Reich DS,Ellisman MH,Rowitch DH,Chan JR,Akassoglou K
Wed Dec 06 00:00:00 UTC 2017
Human Astrocyte Maturation Captured in 3D Cerebral Cortical Spheroids Derived from Pluripotent Stem Cells.
Neuron
Sloan SA,Darmanis S,Huber N,Khan TA,Birey F,Caneda C,Reimer R,Quake SR,Barres BA,Paşca SP
Wed Aug 16 00:00:00 UTC 2017
Discovery of long-range inhibitory signaling to ensure single axon formation.
Nature communications
Takano T,Wu M,Nakamuta S,Naoki H,Ishizawa N,Namba T,Watanabe T,Xu C,Hamaguchi T,Yura Y,Amano M,Hahn KM,Kaibuchi K
Mon Jun 26 00:00:00 UTC 2017
Scalable Production of iPSC-Derived Human Neurons to Identify Tau-Lowering Compounds by High-Content Screening.
Stem cell reports
Wang C,Ward ME,Chen R,Liu K,Tracy TE,Chen X,Xie M,Sohn PD,Ludwig C,Meyer-Franke A,Karch CM,Ding S,Gan L
Tue Oct 10 00:00:00 UTC 2017
Three-dimensional aligned nanofibers-hydrogel scaffold for controlled non-viral drug/gene delivery to direct axon regeneration in spinal cord injury treatment.
Scientific reports
Nguyen LH,Gao M,Lin J,Wu W,Wang J,Chew SY
Tue Feb 07 00:00:00 UTC 2017
LRP1 regulates peroxisome biogenesis and cholesterol homeostasis in oligodendrocytes and is required for proper CNS myelin development and repair.
eLife
Lin JP,Mironova YA,Shrager P,Giger RJ
450-03 was used in Cell Culture to reveal a novel role for low-density lipoprotein receptor-related protein-1 in peroxisome biogenesis, lipid homeostasis, and oligodendrocyte progenitor cell differentiation during white matter development and repair.
Mon Dec 18 00:00:00 UTC 2017
Generation of Mouse Haploid Somatic Cells by Small Molecules for Genome-wide Genetic Screening.
Cell reports
He ZQ,Xia BL,Wang YK,Li J,Feng GH,Zhang LL,Li YH,Wan HF,Li TD,Xu K,Yuan XW,Li YF,Zhang XX,Zhang Y,Wang L,Li W,Zhou Q
450-03 was used in Cell Culture to demonstrate that the spontaneous diploidisation of haploid embryonic stem cells happens in metaphase due to mitotic slippage.
Tue Aug 29 00:00:00 UTC 2017
Generation of Otic Sensory Neurons from Mouse Embryonic Stem Cells in 3D Culture.
Frontiers in cellular neuroscience
Perny M,Ting CC,Kleinlogel S,Senn P,Roccio M
Wed Sep 30 00:00:00 UTC 2020
Defined and Scalable Differentiation of Human Oligodendrocyte Precursors from Pluripotent Stem Cells in a 3D Culture System.
Stem cell reports
Rodrigues GMC,Gaj T,Adil MM,Wahba J,Rao AT,Lorbeer FK,Kulkarni RU,Diogo MM,Cabral JMS,Miller EW,Hockemeyer D,Schaffer DV
Tue Jun 06 00:00:00 UTC 2017
Conditional deletion of L1CAM in human neurons impairs both axonal and dendritic arborization and action potential generation.
The Journal of experimental medicine
Patzke C,Acuna C,Giam LR,Wernig M,Südhof TC
Mon Apr 04 00:00:00 UTC 2016
Prevalent presence of periodic actin-spectrin-based membrane skeleton in a broad range of neuronal cell types and animal species.
Proceedings of the National Academy of Sciences of the United States of America
He J,Zhou R,Wu Z,Carrasco MA,Kurshan PT,Farley JE,Simon DJ,Wang G,Han B,Hao J,Heller E,Freeman MR,Shen K,Maniatis T,Tessier-Lavigne M,Zhuang X
Tue May 24 00:00:00 UTC 2016
Human neural crest cells contribute to coat pigmentation in interspecies chimeras after in utero injection into mouse embryos.
Proceedings of the National Academy of Sciences of the United States of America
Cohen MA,Wert KJ,Goldmann J,Markoulaki S,Buganim Y,Fu D,Jaenisch R
Tue Feb 09 00:00:00 UTC 2016
Generation of highly enriched V2a interneurons from mouse embryonic stem cells.
Experimental neurology
Iyer NR,Huettner JE,Butts JC,Brown CR,Sakiyama-Elbert SE
450-03 was used in Cell Culture to provide a unique tool to begin studying V2a INs in isolation or for use in in vitro models of spinal microcircuits.
Tue Mar 01 00:00:00 UTC 2016
Sustained release of neurotrophin-3 via calcium phosphate-coated sutures promotes axonal regeneration after spinal cord injury.
Journal of neuroscience research
Hanna A,Thompson DL,Hellenbrand DJ,Lee JS,Madura CJ,Wesley MG,Dillon NJ,Sharma T,Enright CJ,Murphy WL
450-03 was used in In vivo experiments to demonstrate that PNGs serve as scaffolds for axonal growth after SCI and that CaP-coated sutures can efficiently release NT-3 to increase axonal regeneration.
Fri Jul 01 00:00:00 UTC 2016
Organ of Corti explants direct tonotopically graded morphology of spiral ganglion neurons in vitro.
The Journal of comparative neurology
Smith FL,Davis RL
450-03 was used in Cell Culture to study the interplay between different regulators of neuronal morphology using cocultured neuron explants with peripheral target tissues removed from distinct cochlear locations.
Mon Aug 01 00:00:00 UTC 2016
Firing frequency and entrainment maintained in primary auditory neurons in the presence of combined BDNF and NT3.
Scientific reports
Wright T,Gillespie LN,O'Leary SJ,Needham K
Thu Jun 23 00:00:00 UTC 2016
Round-window delivery of neurotrophin 3 regenerates cochlear synapses after acoustic overexposure.
Scientific reports
Suzuki J,Corfas G,Liberman MC
Mon Apr 25 00:00:00 UTC 2016
BMS-708163 and Nilotinib restore synaptic dysfunction in human embryonic stem cell-derived Alzheimer's disease models.
Scientific reports
Nishioka H,Tooi N,Isobe T,Nakatsuji N,Aiba K
Mon Sep 19 00:00:00 UTC 2016
Generation and transplantation of reprogrammed human neurons in the brain using 3D microtopographic scaffolds.
Nature communications
Carlson AL,Bennett NK,Francis NL,Halikere A,Clarke S,Moore JC,Hart RP,Paradiso K,Wernig M,Kohn J,Pang ZP,Moghe PV
Thu Mar 17 00:00:00 UTC 2016
Generation and transplantation of reprogrammed human neurons in the brain using 3D microtopographic scaffolds.
Nature communications
Carlson AL,Bennett NK,Francis NL,Halikere A,Clarke S,Moore JC,Hart RP,Paradiso K,Wernig M,Kohn J,Pang ZP,Moghe PV
Thu Mar 17 00:00:00 UTC 2016
Recovery from Toxic-Induced Demyelination Does Not Require the NG2 Proteoglycan.
PloS one
Albrecht S,Hagemeier K,Ehrlich M,Kemming C,Trotter J,Kuhlmann T
450-03 was used in Cell Culture to influence the expression of genes, such as prostaglandin D2 synthase which has neuromodulatory properties.
Fri Nov 17 00:00:00 UTC 2017
Analysis of conditional heterozygous STXBP1 mutations in human neurons.
The Journal of clinical investigation
Patzke C,Han Y,Covy J,Yi F,Maxeiner S,Wernig M,Südhof TC
Tue Sep 01 00:00:00 UTC 2015
Analysis of conditional heterozygous STXBP1 mutations in human neurons.
The Journal of clinical investigation
Patzke C,Han Y,Covy J,Yi F,Maxeiner S,Wernig M,Südhof TC
Tue Sep 01 00:00:00 UTC 2015
MHCII-independent CD4+ T cells protect injured CNS neurons via IL-4.
The Journal of clinical investigation
Walsh JT,Hendrix S,Boato F,Smirnov I,Zheng J,Lukens JR,Gadani S,Hechler D,Gölz G,Rosenberger K,Kammertöns T,Vogt J,Vogelaar C,Siffrin V,Radjavi A,Fernandez-Castaneda A,Gaultier A,Gold R,Kanneganti TD,Nitsch R,Zipp F,Kipnis J
Sun Feb 01 00:00:00 UTC 2015
Excision of Expanded GAA Repeats Alleviates the Molecular Phenotype of Friedreich's Ataxia.
Molecular therapy : the journal of the American Society of Gene Therapy
Li Y,Polak U,Bhalla AD,Rozwadowska N,Butler JS,Lynch DR,Dent SYR,Napierala M
450-03 was used in Cell Culture to investigate GAA repeats in Friedrich’s ataxia.
Mon Jun 01 00:00:00 UTC 2015
Characterization of Nestin-positive stem Leydig cells as a potential source for the treatment of testicular Leydig cell dysfunction.
Cell research
Jiang MH,Cai B,Tuo Y,Wang J,Zang ZJ,Tu X,Gao Y,Su Z,Li W,Li G,Zhang M,Jiao J,Wan Z,Deng C,Lahn BT,Xiang AP
Mon Dec 01 00:00:00 UTC 2014
Human glial chimeric mice reveal astrocytic dependence of JC virus infection.
The Journal of clinical investigation
Kondo Y,Windrem MS,Zou L,Chandler-Militello D,Schanz SJ,Auvergne RM,Betstadt SJ,Harrington AR,Johnson M,Kazarov A,Gorelik L,Goldman SA
Mon Dec 01 00:00:00 UTC 2014
Endothelial NT-3 delivered by vasculature and CSF promotes quiescence of subependymal neural stem cells through nitric oxide induction.
Neuron
Delgado AC,Ferrón SR,Vicente D,Porlan E,Perez-Villalba A,Trujillo CM,D'Ocón P,Fariñas I
Wed Aug 06 00:00:00 UTC 2014
Endothelial NT-3 delivered by vasculature and CSF promotes quiescence of subependymal neural stem cells through nitric oxide induction.
Neuron
Delgado AC,Ferrón SR,Vicente D,Porlan E,Perez-Villalba A,Trujillo CM,D'Ocón P,Fariñas I
Wed Aug 06 00:00:00 UTC 2014
Stretch-activated ion channel Piezo1 directs lineage choice in human neural stem cells.
Proceedings of the National Academy of Sciences of the United States of America
Pathak MM,Nourse JL,Tran T,Hwe J,Arulmoli J,Le DT,Bernardis E,Flanagan LA,Tombola F
Tue Nov 11 00:00:00 UTC 2014
Dysregulation of gene expression as a cause of Cockayne syndrome neurological disease.
Proceedings of the National Academy of Sciences of the United States of America
Wang Y,Chakravarty P,Ranes M,Kelly G,Brooks PJ,Neilan E,Stewart A,Schiavo G,Svejstrup JQ
Tue Oct 07 00:00:00 UTC 2014
Direct conversion of human fibroblasts into neuronal restricted progenitors.
The Journal of biological chemistry
Zou Q,Yan Q,Zhong J,Wang K,Sun H,Yi X,Lai L
Fri Feb 21 00:00:00 UTC 2014
The evolutionary young miR-1290 favors mitotic exit and differentiation of human neural progenitors through altering the cell cycle proteins.
Cell death & disease
Yelamanchili SV,Morsey B,Harrison EB,Rennard DA,Emanuel K,Thapa I,Bastola DR,Fox HS
Thu Jan 09 00:00:00 UTC 2014
Purification of dorsal root ganglion neurons from rat by immunopanning.
Cold Spring Harbor protocols
Zuchero JB
450-03 was used in Sample Preparation to propose a method to study the role of glia in the biology of dorsal root ganglion neurons (DRGs).
Fri Aug 01 00:00:00 UTC 2014
Direct conversion of fibroblasts to neurons by reprogramming PTB-regulated microRNA circuits.
Cell
Xue Y,Ouyang K,Huang J,Zhou Y,Ouyang H,Li H,Wang G,Wu Q,Wei C,Bi Y,Jiang L,Cai Z,Sun H,Zhang K,Zhang Y,Chen J,Fu XD
Thu Jan 17 00:00:00 UTC 2013
Direct conversion of fibroblasts to neurons by reprogramming PTB-regulated microRNA circuits.
Cell
Xue Y,Ouyang K,Huang J,Zhou Y,Ouyang H,Li H,Wang G,Wu Q,Wei C,Bi Y,Jiang L,Cai Z,Sun H,Zhang K,Zhang Y,Chen J,Fu XD
Thu Jan 17 00:00:00 UTC 2013
Rapid single-step induction of functional neurons from human pluripotent stem cells.
Neuron
Zhang Y,Pak C,Han Y,Ahlenius H,Zhang Z,Chanda S,Marro S,Patzke C,Acuna C,Covy J,Xu W,Yang N,Danko T,Chen L,Wernig M,Südhof TC
Wed Jun 05 00:00:00 UTC 2013
Heparin-binding domain of fibrin(ogen) binds growth factors and promotes tissue repair when incorporated within a synthetic matrix.
Proceedings of the National Academy of Sciences of the United States of America
Martino MM,Briquez PS,Ranga A,Lutolf MP,Hubbell JA
Tue Mar 19 00:00:00 UTC 2013
Purification of oligodendrocyte precursor cells from rat cortices by immunopanning.
Cold Spring Harbor protocols
Dugas JC,Emery B
450-03 was used in Cell Culture to demonstrate how to purify oligodendrocyte precursor cells (OPCs) from postnatal rodent brains.
Thu Aug 01 00:00:00 UTC 2013
Long-distance growth and connectivity of neural stem cells after severe spinal cord injury.
Cell
Lu P,Wang Y,Graham L,McHale K,Gao M,Wu D,Brock J,Blesch A,Rosenzweig ES,Havton LA,Zheng B,Conner JM,Marsala M,Tuszynski MH
Fri Sep 14 00:00:00 UTC 2012
Coseeded Schwann cells myelinate neurites from differentiated neural stem cells in neurotrophin-3-loaded PLGA carriers.
International journal of nanomedicine
Xiong Y,Zhu JX,Fang ZY,Zeng CG,Zhang C,Qi GL,Li MH,Zhang W,Quan DP,Wan J
Wed Sep 26 00:00:00 UTC 2012
Coseeded Schwann cells myelinate neurites from differentiated neural stem cells in neurotrophin-3-loaded PLGA carriers.
International journal of nanomedicine
Xiong Y,Zhu JX,Fang ZY,Zeng CG,Zhang C,Qi GL,Li MH,Zhang W,Quan DP,Wan J
Wed Sep 26 00:00:00 UTC 2012
miR-200c targets a NF-κB up-regulated TrkB/NTF3 autocrine signaling loop to enhance anoikis sensitivity in triple negative breast cancer.
PloS one
Howe EN,Cochrane DR,Cittelly DM,Richer JK
Thu May 30 00:00:00 UTC 2013
Oligodendrocyte differentiation from adult multipotent stem cells is modulated by glutamate.
Cell death & disease
Cavaliere F,Urra O,Alberdi E,Matute C
Thu Feb 02 00:00:00 UTC 2012
3D multicolor super-resolution imaging offers improved accuracy in neuron tracing.
PloS one
Lakadamyali M,Babcock H,Bates M,Zhuang X,Lichtman J
Mon Jun 04 00:00:00 UTC 2012
MicroRNA-mediated conversion of human fibroblasts to neurons.
Nature
Yoo AS,Sun AX,Li L,Shcheglovitov A,Portmann T,Li Y,Lee-Messer C,Dolmetsch RE,Tsien RW,Crabtree GR
Wed Jul 13 00:00:00 UTC 2011
Directed conversion of Alzheimer's disease patient skin fibroblasts into functional neurons.
Cell
Qiang L,Fujita R,Yamashita T,Angulo S,Rhinn H,Rhee D,Doege C,Chau L,Aubry L,Vanti WB,Moreno H,Abeliovich A
Fri Aug 05 00:00:00 UTC 2011
Identification of a low-molecular weight TrkB antagonist with anxiolytic and antidepressant activity in mice.
The Journal of clinical investigation
Cazorla M,Prémont J,Mann A,Girard N,Kellendonk C,Rognan D
Sun May 01 00:00:00 UTC 2011
Astrocytes carrying the superoxide dismutase 1 (SOD1G93A) mutation induce wild-type motor neuron degeneration in vivo.
Proceedings of the National Academy of Sciences of the United States of America
Papadeas ST,Kraig SE,O'Banion C,Lepore AC,Maragakis NJ
Tue Oct 25 00:00:00 UTC 2011
The PI3K-Akt pathway inhibits senescence and promotes self-renewal of human skin-derived precursors in vitro.
Aging cell
Liu S,Liu S,Wang X,Zhou J,Cao Y,Wang F,Duan E
Mon Aug 01 00:00:00 UTC 2011
Biophysical characteristics reveal neural stem cell differentiation potential.
PloS one
Labeed FH,Lu J,Mulhall HJ,Marchenko SA,Hoettges KF,Estrada LC,Lee AP,Hughes MP,Flanagan LA
Thu Jan 26 00:00:00 UTC 2012
Canonical Wnt signalling requires the BMP pathway to inhibit oligodendrocyte maturation.
ASN neuro
Feigenson K,Reid M,See J,Crenshaw III EB,Grinspan JB
Thu Jun 16 00:00:00 UTC 2011
Canonical Wnt signalling requires the BMP pathway to inhibit oligodendrocyte maturation.
ASN neuro
Feigenson K,Reid M,See J,Crenshaw III EB,Grinspan JB
Thu Jun 16 00:00:00 UTC 2011
Efficient derivation of human neuronal progenitors and neurons from pluripotent human embryonic stem cells with small molecule induction.
Journal of visualized experiments : JoVE
Parsons XH,Teng YD,Parsons JF,Snyder EY,Smotrich DB,Moore DA
Fri Oct 28 00:00:00 UTC 2011
Gas6 increases myelination by oligodendrocytes and its deficiency delays recovery following cuprizone-induced demyelination.
PloS one
Binder MD,Xiao J,Kemper D,Ma GZ,Murray SS,Kilpatrick TJ
Thu Mar 10 00:00:00 UTC 2011
Derivation, propagation and controlled differentiation of human embryonic stem cells in suspension.
Nature biotechnology
Steiner D,Khaner H,Cohen M,Even-Ram S,Gil Y,Itsykson P,Turetsky T,Idelson M,Aizenman E,Ram R,Berman-Zaken Y,Reubinoff B
Thu Apr 01 00:00:00 UTC 2010
The Angelman Syndrome protein Ube3A regulates synapse development by ubiquitinating arc.
Cell
Greer PL,Hanayama R,Bloodgood BL,Mardinly AR,Lipton DM,Flavell SW,Kim TK,Griffith EC,Waldon Z,Maehr R,Ploegh HL,Chowdhury S,Worley PF,Steen J,Greenberg ME
Fri Mar 05 00:00:00 UTC 2010
Downstream of tyrosine kinase/docking protein 6, as a novel substrate of tropomyosin-related kinase C receptor, is involved in neurotrophin 3-mediated neurite outgrowth in mouse cortex neurons.
BMC biology
Li Wq,Shi L,You Yg,Gong Yh,Yin B,Yuan Jg,Peng Xz
Fri Jun 18 00:00:00 UTC 2010
Downstream of tyrosine kinase/docking protein 6, as a novel substrate of tropomyosin-related kinase C receptor, is involved in neurotrophin 3-mediated neurite outgrowth in mouse cortex neurons.
BMC biology
Li Wq,Shi L,You Yg,Gong Yh,Yin B,Yuan Jg,Peng Xz
Fri Jun 18 00:00:00 UTC 2010
Kinetic analysis of neurotrophin-3-mediated differentiation of embryonic stem cells into neurons.
Tissue engineering. Part A
Willerth SM,Sakiyama-Elbert SE
Sun Feb 01 00:00:00 UTC 2009
Dendritic branching of olfactory bulb mitral and tufted cells: regulation by TrkB.
PloS one
Imamura F,Greer CA
Tue Aug 25 00:00:00 UTC 2009
Disorganized innervation and neuronal loss in the inner ear of Slitrk6-deficient mice.
PloS one
Katayama K,Zine A,Ota M,Matsumoto Y,Inoue T,Fritzsch B,Aruga J
Wed Nov 11 00:00:00 UTC 2009
Activation of Trk neurotrophin receptors by glucocorticoids provides a neuroprotective effect.
Proceedings of the National Academy of Sciences of the United States of America
Jeanneteau F,Garabedian MJ,Chao MV
Tue Mar 25 00:00:00 UTC 2008
Isolation of neural stem and precursor cells from rodent tissue.
Methods in molecular biology (Clifton, N.J.)
Wu Y,Liu Y,Chesnut JD,Rao MS
450-03 was used in Cell Culture to provide a protocol for dissection of rodent embryonic spinal cords by enzymatic separation, and isolation and enrichment (or purification) of neuronal and glial precursors at different developing stages by fluorescence-activated cell sorting.
Thu Apr 17 00:00:00 UTC 2008
Effects of NGF, NT-3 and GDNF family members on neurite outgrowth and migration from pelvic ganglia from embryonic and newborn mice.
BMC developmental biology
Stewart AL,Anderson RB,Kobayashi K,Young HM
Fri Jul 25 00:00:00 UTC 2008
Fgfr2b mediated epithelial-mesenchymal interactions coordinate tooth morphogenesis and dental trigeminal axon patterning.
Mechanisms of development
Kettunen P,Spencer-Dene B,Furmanek T,Kvinnsland IH,Dickson C,Thesleff I,Luukko K
Wed Mar 19 00:00:00 UTC 2008
Regulation of apoptosis and differentiation by p53 in human embryonic stem cells.
The Journal of biological chemistry
Qin H,Yu T,Qing T,Liu Y,Zhao Y,Cai J,Li J,Song Z,Qu X,Zhou P,Wu J,Ding M,Deng H
Fri Feb 23 00:00:00 UTC 2007
Neurotrophins mediate human embryonic stem cell survival.
Nature biotechnology
Pyle AD,Lock LF,Donovan PJ
Wed Mar 01 00:00:00 UTC 2006
Protection by GDNF and other trophic factors against the dopamine-depleting effects of neurotoxic doses of methamphetamine.
Annals of the New York Academy of Sciences
Cass WA,Peters LE,Harned ME,Seroogy KB
Tue Aug 01 00:00:00 UTC 2006
Side population cells isolated from porcine dental pulp tissue with self-renewal and multipotency for dentinogenesis, chondrogenesis, adipogenesis, and neurogenesis.
Stem cells (Dayton, Ohio)
Iohara K,Zheng L,Ito M,Tomokiyo A,Matsushita K,Nakashima M
Wed Nov 01 00:00:00 UTC 2006
Side population cells isolated from porcine dental pulp tissue with self-renewal and multipotency for dentinogenesis, chondrogenesis, adipogenesis, and neurogenesis.
Stem cells (Dayton, Ohio)
Iohara K,Zheng L,Ito M,Tomokiyo A,Matsushita K,Nakashima M
Wed Nov 01 00:00:00 UTC 2006
Side population cells isolated from porcine dental pulp tissue with self-renewal and multipotency for dentinogenesis, chondrogenesis, adipogenesis, and neurogenesis.
Stem cells (Dayton, Ohio)
Iohara K,Zheng L,Ito M,Tomokiyo A,Matsushita K,Nakashima M
Wed Nov 01 00:00:00 UTC 2006
Side population cells isolated from porcine dental pulp tissue with self-renewal and multipotency for dentinogenesis, chondrogenesis, adipogenesis, and neurogenesis.
Stem cells (Dayton, Ohio)
Iohara K,Zheng L,Ito M,Tomokiyo A,Matsushita K,Nakashima M
Wed Nov 01 00:00:00 UTC 2006
Generation and characterization of oligodendrocytes from lineage-selectable embryonic stem cells in vitro.
Methods in molecular biology (Clifton, N.J.)
Billon N,Jolicoeur C,Raff M
450-03 was used in Cell Culture to culture and efficently transform modified mouse ES cells to neural precursor cells, OPCs or oligodendrocytes.
Tue Aug 22 00:00:00 UTC 2006
Exploring the regulation of human neural precursor cell differentiation using arrays of signaling microenvironments.
Molecular systems biology
Soen Y,Mori A,Palmer TD,Brown PO
Fri Sep 29 00:00:00 UTC 2006
Neurotrophins induce formation of functional excitatory and inhibitory synapses between cultured hippocampal neurons.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Vicario-Abejón C,Collin C,McKay RD,Segal M
Tue Sep 15 00:00:00 UTC 1998
Regulation of quantal secretion by neurotrophic factors at developing motoneurons in Xenopus cell cultures.
The Journal of physiology
Liou JC,Yang RS,Fu WM
Fri Aug 15 00:00:00 UTC 1997