Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Highest antigen sequence indentity to the following orthologs: Mouse (75%), Rat (75%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65292. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
PLCB2 is the production of the second messenger molecules diacylglycerol (DAG) and inositol 1,4,5-trisphosphate (IP3) is mediated by activated phosphatidylinositol-specific phospholipase C enzymes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-2; Phosphoinositide phospholipase C-beta-2; phospholipase C, beta 2; Phospholipase C-beta-2; PLC beta-2; PLC-beta-2; PLC-beta2; PLCbeta-2; PLCbeta2
Gene Aliases: PLC-beta-2; PLCB2
UniProt ID: (Human) Q00722
Entrez Gene ID: (Human) 5330
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support