Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This peptide corresponds to 13 amino acids near the carboxy terminus of human PPAPDC1A.
PEP-0978 can be used as a blocking peptide with polyclonal antibody PA5-20864.
Phosphatidate phosphatase (PAP) plays important role in lipid-signaling metabolism in eukaryotic cells. Two distinct types of PAP (PAP1 and PAP2) activity have been distinguished by their subcellular localization and differential sensitivity to N-ethylmaleimide(NEM) and Mg2+. A yeast diacylglycerol pyrophosphate (DGPP) phosphatase (DPP1) and mammalian DGPP phosphatase (PAP2) have been identified as Mg2+-independent and NEM-insensitive membrane-associated. PPAPDC1A (also known as DPPL2) and PPAPDC1B (DPPL1) form a novel type of Mg2+-independent and NEM-sensitive mammalian phosphatidate phosphatase showing broad substrate specificity. PPAPDC1A is preferentially expressed in endothelial cells. Studies of PPAPDC1A and PAP activity suggest that they may play a role in angiogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: diacylglycerol pyrophosphate like 2; diacylglycerol pyrophosphate phosphatase-like 2; phosphatidate phosphatase PPAPDC1A; phosphatidic acid phosphatase type 2 domain containing 1A; Phosphatidic acid phosphatase type 2 domain-containing protein 1A; Phospholipid phosphatase 4; RP11-257O17.1
Gene Aliases: DPPL2; PLPP4; PPAPDC1; PPAPDC1A
UniProt ID: (Human) Q5VZY2
Entrez Gene ID: (Human) 196051
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support