Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This peptide corresponds to 20 amino acids near the amino terminus of human TP1
PEP-0474 can be used as a blocking peptide with polyclonal antibody PA5-20354.
Telomerase is an RNA-dependent DNA polymerase that uses an RNA component to add telomeric repeat sequences at the ends of chromosomes. Besides the RNA component which serves as the template that specifies the telomeric repeat, the telomerase complex contains a reverse transcriptase protein (TRT) and various accessory proteins including the telomerase-associated protein 1 (TP1). Telomerase activity is low in most somatic cells, causing the gradual shortening of telomeres which can ultimately lead to telomere fusion and cell death. High levels of telomerase activity are widely seen in cancerous cells and while recent experiments have suggested that telomerase may be a viable target in cancer therapy, expression levels of TP1 do not correlate with malignancy. At least two isoforms of TP1 are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: p240; p80 telomerase homolog; Telomerase protein 1; Telomerase protein component 1; Telomerase-associated protein 1; TROVE domain family, member 1
Gene Aliases: p240; TEP1; TLP1; TP1; TROVE1; VAULT2
UniProt ID: (Human) Q99973
Entrez Gene ID: (Human) 7011
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support