Invitrogen antibodies—maximize lab budgets with advanced validation & coverage

Shop antibodies

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
  • Special Offers
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Proteins & Peptides
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Proteins & Peptides ›
            • TGF beta-1 Proteins
            Activate Wnt signaling with improved solubility. Try Gibco Wnt Surrogate-Fc Fusion Protein

            Gibco

            Human TGF-beta 1 Recombinant Protein

            5 References
            View all (9) TGF beta-1 proteins
            Datasheet
            Tech Support
            Datasheet
            Tech Support

            Cite Human TGF-beta 1 Recombinant Protein

            • Testing Data (3)
            Application
            Human TGF-beta 1 Protein in Functional Assay (Functional)
            Group 53 Created with Sketch.

            FIGURE: 1 / 3

            Human TGF-beta 1 Protein (PHG9214) in Functional

            Immunofluorescence analysis of catenin beta-1 (CTNNB1) was performed using MCF7 cells treated with 10 ng/mL of human TGF-beta 1 recombinant protein (Product # PHG9214) for 24 hours. The cells were labeled with anti-beta Catenin recombinant rabbit monoclonal antibody (Product # MA5-29202) (Panel a & c;Green). The composite (b & d) represents the merged image with DAPI (blue) and F-actin (red) staining. Cel... View More
            View Product

            Human TGF-beta 1 Protein in Functional Assay (Functional)
            Human TGF-beta 1 Protein in Functional Assay (Functional)
            Human TGF-beta 1 Protein in Functional Assay (Functional)

            Product Details

            PHG9214

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            Assay-dependent
            -

            Immunohistochemistry (IHC)

            Assay-dependent
            -

            ELISA (ELISA)

            Assay-dependent
            -

            Functional Assay (Functional)

            Assay-dependent
            View 5 publications 5 publications

            Control (Ctrl)

            Assay-dependent
            -
            Product Specifications

            Species

            Human

            Published species

            Human, Rat

            Expression System

            HEK293 cells

            Amino acid sequence

            Human TGF-beta 1 recombinant protein is a homodimer containing two 112 amino acid subunits

            Molecular weight

            25 kDa

            Class

            Recombinant

            Type

            Protein

            Purity

            ≥95% by SDS-PAGE

            Endotoxin concentration

            <0.1 ng/µg

            Activity

            ED50 < 0.2 ng/mL; determined by the dose-dependent inhibition of mouse HT-2 cells stimulated with mouse IL-4.

            Conjugate

            Unconjugated

            Form

            Lyophilized

            Purification

            purified

            Storage buffer

            100mM acetic acid

            Contains

            no preservative

            Storage conditions

            4°C

            Shipping conditions

            Wet ice

            Product Specific Information

            Reconstitution: Centrifuge the vial briefly, before opening to bring the contents to the bottom. Reconstitute the lyophilized protein in an acidic buffer, such as 40 mM acetic acid or 4 mM HCl, to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For long term storage, it is recommended to further dilute in buffer containing a carrier protein, such as 0.1% BSA, and store in working aliquots at -20°C to -80°C.

            Storage: Store the lyophilized protein at 2-8°C or -20°C, preferably desiccated. Store reconstituted human TGF-beta 1 at ≤-20°C (not in a frost-free freezer). Avoid repeated freeze-thaw cycles.

            Target Information

            TGF beta-1 (TGFB1, Transforming Growth Factor Beta 1) is a polypeptide member of the transforming growth factor beta superfamily of cytokines, found almost ubiquitously in tissues. Transforming growth factor (TGF)-b is stored in the extracellular matrix as a latent complex with its pro-domain. Activation of TGF beta-1 requires the binding of aV integrin to an RGD sequence in the prodomain and exertion of force on this domain, which is held in the extracellular matrix by latent TGF-b binding proteins. Latent forms are complexes of TGF beta-1, an amino-terminal portion of the TGF-beta precursor, designated TGF-LAP (TGF-latency associated peptide), and a specific binding protein, known as LTBP. TGF beta-1 helps regulates proliferation, differentiation, adhesion, migration in many cell types. Many cells have TGF beta receptors, and the protein positively and negatively regulates many other growth factors. TGF beta-1 is cleaved into a latency-associated peptide and a mature TGF beta-1 peptide, and is found in either a latent form composed of a TGFB1 homodimer, a LAP homodimer, and a latent TGFB1-binding protein, or in an active form composed of a TGF beta-1 homodimer. The mature peptide may also form heterodimers with other TGF beta family members. The gene for TGF beta-1 is frequently upregulated in tumor cells, and mutations in this gene result in Camurati-Engelmann disease and cystic fibrosis.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: latency-associated peptide; prepro-transforming growth factor beta-1; TGF b; TGF b 1; TGF beta; tgf beta 1; TGF beta-1; TGF beta-1 protein; tgf beta1; TGF β; TGF β 1; TGF β1; TGF-B1; TGF-beta 1 protein; TGF-beta-1; TGFβ1; Transforming growth factor; transforming growth factor b1; transforming growth factor beta; Transforming growth factor beta-1 proprotein; transforming growth factor β1

            View more View less

            Gene Aliases: CED; DPD1; LAP; TGFB; TGFB1; TGFbeta

            View more View less

            UniProt ID: (Human) P01137

            View more View less

            Entrez Gene ID: (Human) 7040

            View more View less

            It has to be done as per old AB suggested Products section.
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-6ff95d844f-9jzxh:80/100.66.79.106:80.
            git-commit: f44aa9153aef922f8fad9a05e87b8aee811da128
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.28.0-Offline