Invitrogen antibodies—maximize lab budgets with advanced validation & coverage

Shop antibodies

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Proteins & Peptides
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Proteins & Peptides ›
            • TGF beta-3 Proteins

            Gibco

            Human TGF-beta 3 Recombinant Protein

            View all (5) TGF beta-3 proteins
            Datasheet
            Tech Support
            Promotion Available
            Learn more
            Catalog # PHG9302
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            10,990.00
             
            Check your price
            Request bulk or custom format
            Datasheet
            Tech Support

            Cite Human TGF-beta 3 Recombinant Protein

            Product Details

            PHG9302

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            Assay-dependent
            -

            Immunohistochemistry (IHC)

            Assay-dependent
            -

            ELISA (ELISA)

            Assay-dependent
            -

            Functional Assay (Functional)

            Assay-dependent
            -

            Control (Ctrl)

            Assay-dependent
            -
            Product Specifications

            Species

            Human

            Expression System

            E. coli

            Amino acid sequence

            Human TGF-beta 3 recombinant protein is a homodimer containing two 112 amino acid subunits

            Molecular weight

            25.4 kDa

            Class

            Recombinant

            Type

            Protein

            Purity

            >98% by SDS-PAGE and HPLC

            Endotoxin concentration

            <0.1 ng/µg

            Activity

            ED50 ≤ 0.05 ng/mL (≥ 12.5x10e6 IU/mg); determined by a toxicity assay using the WHO Standard 98/608 for direct comparison.

            Conjugate

            Unconjugated

            Form

            Liquid

            Purification

            purified

            Storage buffer

            0.12% acetic acid with 20% ethanol

            Contains

            no preservative

            Storage conditions

            4°C

            Shipping conditions

            Wet ice (domestic); Wet or Dry ice (international)

            Product Specific Information

            •Purity: >98% by SDS-PAGE and HPLC
            •Endotoxin Concentration: <0.1 ng/µg
            •Protein Length: 112 aa (2 identical subunits)
            •Biological Activity: ED50 ≤0.05 ng/mL
            •Molecular Weight: 17.5 kDa

            Target Information

            Transforming growth factor beta 3 (TGF beta 3) is the third member of the transforming growth factor family of cytokines, which also includes TGF beta 1 and beta 2. These cytokines are secreted in precursor form consisting of a bioactive C-terminal domain attached to an N-terminal domain known as latency associated protein (LAP). Cleavage of LAP results in the mature protein, which functions as a disulfide-linked homodimer. As with all members of the family, TGF beta 3 is highly conserved across species, with mouse and human TGF beta 3 demonstrating 100% sequence homology and cross-species activity.The members of this family can be expressed by most cell types and exert pleiotropic effects, which include the suppression of B- and T-cell effector activity, mediation of tissue healing, and suppression of tumor proliferation. The promotion of CD4+CD25+ T-cell expansion is a newly discovered function of the TGF beta cytokines, and indicates an important role in the protection against autoimmunity.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: FLJ16571; H-TGF-b-3; prepro-transforming growth factor beta-3; tgf beta 3; TGF-beta-3; Transforming growth factor; Transforming growth factor beta-3 proprotein; transforming growth factor, beta 3

            View more View less

            Gene Aliases: ARVD; ARVD1; LDS5; RNHF; TGF-beta3; TGFB3

            View more View less

            UniProt ID: (Human) P10600

            View more View less

            Entrez Gene ID: (Human) 7043

            View more View less

            It has to be done as per old AB suggested Products section.
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-6ff95d844f-mkdxq:80/100.66.77.154:80.
            git-commit: f44aa9153aef922f8fad9a05e87b8aee811da128
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.28.0-Offline