Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66391. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Involved in trafficking and recycling of synaptic vesicles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Transmembrane protein 230; UPF0414 transmembrane protein C20orf30
Gene Aliases: C20orf30; dJ1116H23.2.1; HSPC274; TMEM230; UNQ2432/PRO4992
UniProt ID: (Human) Q0VGC8
Entrez Gene ID: (Human) 29058
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support