Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This peptide corresponds to 14 amino acids near the center of human ZBTB2.
PEP-0779 can be used as a blocking peptide with polyclonal antibody PA5-20659.
ZBTB2 may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger and BTB domain-containing protein 2; ZNF909
Gene Aliases: KIAA1483; ZBTB2; ZNF437
UniProt ID: (Human) Q8N680
Entrez Gene ID: (Human) 57621
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support