Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Highest antigen sequence indentity to the following orthologs: Mouse (29%), Rat (29%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63118. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Transcriptional repressor that plays a role in cell proliferation. Requires TRIM28 for its activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BMZF-3; Bone marrow zinc finger 3; Zinc finger protein 256
Gene Aliases: BMZF-3; BMZF3; ZNF256
UniProt ID: (Human) Q9Y2P7
Entrez Gene ID: (Human) 10172
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support