Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PEP-168 is a 10 amino acid synthetic peptide whose sequence corresponds to GnRH. The sequence of this peptide is (amino to carboxy terminus): pyroE-H-W-S-Y-G-L-R-P- (G-NH2)
This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and GnRH, catalog # PA1-120. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (~70-fold) for competitive inhibition of antibody-protein binding reactions.
Reconstitute with 0.1 mL of distilled water.
The Luteinizing Hormone Releasing Hormone (LH-RH) is produced in the hypothalamus and influences the release of Luteinizing Hormone in the pituitary gland. LH-RH is also known as Gonadotropin-releasing hormone (Gn-RH).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gonadotropin releasing hormone 2; hypogonadal; luteinizing hormone releasing hormone; luteinizing hormone-releasing hormone I; Lutropin; Progonadoliberin I; Progonadoliberin-1
Gene Aliases: Gnrh; Gnrh1; Gnrh2; hpg; LHRH; Lhrh1; Lnrh
UniProt ID: (Mouse) P13562
Entrez Gene ID: (Mouse) 14714
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support