Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
It is recommended that the protein be aliquoted for optimal storage.
It is recommended that sterile water (80 µL) be added to the vial to prepare a stock solution of 0.25 mg/mL.
Concentration is measured by UV.
Hippostasin, also known as KLK11, is a kallikrein-like serine protease, which has two alternatively spliced isoforms, brain-type and prostate-type. Hippostasin plays a role in the prostate, including reproductive and/or tumorigenic functions. Elevated serum levels of KLK11 were found in 70% of women with ovarian cancer and in 60% of men with prostate cancer. Analysis of the KLK11 biomarker in serum may aid in the diagnosis and monitoring of ovarian and prostatic carcinoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hippostasin; kallikrein 11; Kallikrein-11; Kallikrein11; protease, serine, 20; Serine protease 20
Gene Aliases: 2310015I08Rik; Klk11; Prss20; TLSP
UniProt ID: (Mouse) Q9QYN3
Entrez Gene ID: (Mouse) 56538
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support