When it comes to food safety and quality testing for unwanted residues, contaminants, adulteration and more, our start-to-finish workflows enable regulatory and food and beverage analysis laboratories to meet the challenges of today and prepare for the requirements of tomorrow. Supplied in partnership with Health in Code, S.L., a leader in GMO research and development, our solutions are underpinned by market-leading molecular instrumentation for performance you can rely on. 
DNA sample preparation
Whatever your sample throughput, our PCR workflows for food profiling and authenticity begin with our streamlined, scalable DNA extraction solutions. Choose from manual and automated extraction options to suit your lab setup. 
GMO Extraction Kit
GMO Extraction Kit
The Thermo Scientific GMO Extraction Kit enables high-quality, fast and easy silica-based DNA extraction and purification from a wide variety of samples, including raw materials and processed foods.
Throughput: Low to medium (manual)
KingFisher Flex Purification System
Thermo Scientific KingFisher Flex Purification System
Carry out automated total nucleic acid purification and concentration of selected targets from a broad range of food and environmental samples with the Thermo Scientific KingFisher Flex Purification System. Ideal for higher throughput labs, the system, in combination with the Applied Biosystems PrepSEQ Nucleic Acid Extraction Kit, offers a fast solution with reduced hands-on time.
Throughput: medium to higher (automated)
Detection and quantification of targeted meat and fish species
Detection and quantification of targeted meat and fish species
In collaboration with Health in Code, S.L., we offer a broad portfolio of real-time PCR-based species detection and quantification solutions.
GMO screening and quantification
GMO screening and quantification
Choose from GMO screening and quantification solutions that can detect the presence of GMO-specific DNA events in soy, corn, and downstream processed products. We also offer a universal assay that detects and quantifies all approved GMOs in Europe and the vast majority of GMOs approved in other countries.
Detection of animal DNA for ethical food claims and labelling
Detection of animal DNA for ethical food claims and labelling
Evidence your ethical food claims and meet labeling requirements with vegan and halal PCR kits. Simply and reliably detect the presence of animal DNA in raw and processed food samples with a sensitivity down to 0.0005% pork DNA for halal and 0.01% bovine DNA for vegan.
Instrumentation
Effortless, rapid food testing with the QuantStudio 5 Food Safety PCR System
The Applied Biosystems QuantStudio 5 Food Safety Real-Time PCR System is a high-performance benchtop platform, combining ease of use and system connectivity with powerful options that help enable maximum data utility and control. Benefit from the flexibility of a 6-channel, 96-well, cloud-enabled open platform suitable for running our full range of PCR solutions for food safety, authenticity, and quality testing.
For Research Use Only. Not for use in diagnostic procedures.