Images can aid in understanding cell structure and mechanisms.
As a cell biologist, you know how important it is to understand the intricate mechanisms of cell structure and function. However, analyzing data from various imaging systems and modalities that use different file formats can be a daunting task. Each experimental setup presents a unique challenge, and multi-scale dynamic processes require the tracking of objects of various sizes, from diffraction-limited particles to entire cells. With data sets containing anywhere from a few dozen to tens of thousands of objects, identifying and quantifying sub-cellular structures that support intracellular processes can seem overwhelming.
Thermo Scientific Amira Software is a powerful toolbox that enables cell biologists to visualize, process, and analyze complex biological data in 3D. With its advanced image processing, visualization, and analysis tools, Amira software can help you gain insights into the structure and function of cells and tissues that would be impossible to see with 2D images alone.
Whether you're studying cellular processes, tissue organization, or the effects of drugs on cells, Amira software can help you gain a deeper understanding of your images. With our easy-to-use interface and comprehensive tools, you can streamline your workflow and spend more time doing what you do best - advancing the field of cell biology. 
Video Player is loading.
Current Time 0:00
Duration 1:36
Loaded: 10.15%
Stream Type LIVE
Remaining Time 1:36
1x
    • Chapters
    • descriptions off, selected
    • en (Main), selected
    Imaging data
    Workflow using Avizo Software
    Processing
    Workflow using Avizo Software
    Visualization
    Workflow using Avizo Software
    Analysis
    Workflow using Avizo Software
    Customization
    Confidence
    Thanks to powerful segmentation and image processing capabilities and workflows, and after 20+ years of collaboration with the scientific community and thousands of researchers, it has been now proven that our digital imaging-based workflows provide reliable answers to biomedical and life science research.
    Customization
    Customization
    Because your needs are unique and keep evolving, our software solutions are fully flexible and open to customization. Thanks to our scripting interface (Python, TCL), bridge with MATLAB, and our programming API, you can expand our software solution and integrate your own IP (Intellectual Property). And, if needed, our professional service team can help you design unique solutions tailored to your needs.
    icon-improve_collaboration_64x64
    Support
    Thanks to our dedicated professional support team, you get access to our top experts to ensure that no question is left unanswered. And with our training, consulting options, and ever-expanding collection of tutorials and how-to’s, you can reduce your learning curve and focus on getting the answers you are pursuing.
    Software_Automation_65x64
    Automation
    Thanks to our automation capabilities, and with the addition of our always expanding online repository of add-ons (Xtra Gallery), you can encapsulate repeatable workflows into easy-to-reproduce recipes. With the addition of artificial intelligence, analysis can be performed by non-image processing experts, allowing them to save time on complex analysis while ensuring results consistency.
    Introduction to Amira Software for cell biology
    During this 20-minutes webinar, you will learn how to:
    • Accelerate subcellular (membrane, organelle) detection and segmentation from Electron Microscopy (TEM) data using a range of readily available tools, including 3D Deep Learning model training.
    • Analyze multichannel fluorescence endothelial cell culture data by performing fast and effective cleaning, segmentation, volume colocalization and neighbor cell proximity.
    • Extract complex information in 3D such as detection, segmentation and penetration of cancer or immune cells to manage multichannel High Content Screening multi-well images of organoid models.
    • Quickly convert, visualize and accurately process large (~100GB) multi-channel and time series light sheet tissue imaging data.
    Use cases and insight from your peers
    Explore industry-leading insights and research that can support your lab workflow.
    Helping scientists answer questions that enable breakthrough discoveries in life sciences, materials science, and industry.
    Correlative Microscopy: Using Amira Software to Understand the Spread of Cancer
    Read more
    Amira Software Accelerates Veterinary Research with 3D Visualization and Analysis
    Read more
    Molecular Clarity—Discovering What’s Possible with Cryo-Electron Tomography
    Read more
    Cryo-Electron Tomography and 3D Software Advances Coronavirus Research
    Read more
    Register now to get exclusive content delivered directly to your inbox.
    Register
    Techniques
    Cell detection
    Cellular spheroids are 3D cell cultures that represent in vivo tissue better than traditional cellular monolayers, improving the ability of researchers to study cancer, diabetes mellitus, or stem cells, among many applications. However, cell analysis in such 3D multicellular objects is not accurately possible, especially without destroying the object of study. Amira Software can automatically separate the cells and perform volumetric, morphometric, and intensity measurements on individual cells and their nuclei in 3D image data of these spheroids. Amira Software also helps with the analysis of conventional 2D cellular monolayers.
    Correlative imaging
    With its long-standing applications in multi-modality imaging techniques, Amira Software is the tool of choice for many researchers working with data from different imaging modalities. Amira Software’s architecture supports multi-volume data processing and visualization from the ground up. The automatic registration tools integrated into the standard edition of Amira Software with the Multi-Planar Workroom offer the flexibility of the most commonly used automated registration metrics and optimizers that allow for the adjustment of the registration parameters to meet the needs of your data.
    Membrane detection
    Amira Software accelerates segmentation by employing automated detection of cellular features, such as filaments and membranes. By performing advanced image analysis and statistics on the cryo-tomograms, Amira Software combines data from these individual structures, averaging out noise and increasing contrast to create a composite model with higher resolution.
    Multi-channel and time-series visualization
    With our new Xplore5D extension for Amira Software, life scientists can easily visualize, correlate, and process large multichannel and time series data in one platform, without worrying about image size or memory limits. With its smart file format, Xplore5D allows you to compress all imaging data without losing precious information collected during acquisition.
    Learn more
    High-content screening
    Amira Software enables multi-channel quantitative capabilities to investigate the most demanding scientific and experimental models used in HCS, including immuno-oncology, neuroscience, and organoid health. Thanks to its incredible speed and flexibility, Amira Software helps enable advanced 2D–5D bioimaging workflows in research areas ranging from structural and cellular biology to neuronal and spheroid morphology in 3D models. To facilitate these advanced analyses, Amira Software features deep learning for improved training and prediction, including "intelligent denoising functionality.
    Learn more
    Filament tracing
    With Amira Software, you are able to use a template-matching algorithm to automatically detect and trace these fine filaments in noisy cryo-EM or DualBeam instrument data. In addition, Amira Software enables you to reconstruct filamentous networks and edit the resulting graphs to remove image features erroneously identified as filaments or to add missing parts of a network. Graphs of filaments can then be quantified by measuring parameters such as amount, length, thickness, orientation, ranks, etc. Compelling 3D renderings can be created from the reconstructed model and the computed parameters.
    Cell detection
    Cellular spheroids are 3D cell cultures that represent in vivo tissue better than traditional cellular monolayers, improving the ability of researchers to study cancer, diabetes mellitus, or stem cells, among many applications. However, cell analysis in such 3D multicellular objects is not accurately possible, especially without destroying the object of study. Amira Software can automatically separate the cells and perform volumetric, morphometric, and intensity measurements on individual cells and their nuclei in 3D image data of these spheroids. Amira Software also helps with the analysis of conventional 2D cellular monolayers.
    Correlative imaging
    With its long-standing applications in multi-modality imaging techniques, Amira Software is the tool of choice for many researchers working with data from different imaging modalities. Amira Software’s architecture supports multi-volume data processing and visualization from the ground up. The automatic registration tools integrated into the standard edition of Amira Software with the Multi-Planar Workroom offer the flexibility of the most commonly used automated registration metrics and optimizers that allow for the adjustment of the registration parameters to meet the needs of your data.
    Membrane detection
    Amira Software accelerates segmentation by employing automated detection of cellular features, such as filaments and membranes. By performing advanced image analysis and statistics on the cryo-tomograms, Amira Software combines data from these individual structures, averaging out noise and increasing contrast to create a composite model with higher resolution.
    Multi-channel and time-series visualization
    With our new Xplore5D extension for Amira Software, life scientists can easily visualize, correlate, and process large multichannel and time series data in one platform, without worrying about image size or memory limits. With its smart file format, Xplore5D allows you to compress all imaging data without losing precious information collected during acquisition.
    Learn more
    High-content screening
    Amira Software enables multi-channel quantitative capabilities to investigate the most demanding scientific and experimental models used in HCS, including immuno-oncology, neuroscience, and organoid health. Thanks to its incredible speed and flexibility, Amira Software helps enable advanced 2D–5D bioimaging workflows in research areas ranging from structural and cellular biology to neuronal and spheroid morphology in 3D models. To facilitate these advanced analyses, Amira Software features deep learning for improved training and prediction, including "intelligent denoising functionality.
    Learn more
    Filament tracing
    With Amira Software, you are able to use a template-matching algorithm to automatically detect and trace these fine filaments in noisy cryo-EM or DualBeam instrument data. In addition, Amira Software enables you to reconstruct filamentous networks and edit the resulting graphs to remove image features erroneously identified as filaments or to add missing parts of a network. Graphs of filaments can then be quantified by measuring parameters such as amount, length, thickness, orientation, ranks, etc. Compelling 3D renderings can be created from the reconstructed model and the computed parameters.
    Services
    Amira-Avizo-Introductory-training_1160x600
    Introductory training
    Shorten your learning curve and maximize your investment with this introductory training specifically designed for new users of Amira, Avizo and PerGeos Software.
    The course consists of a lecture with hands-on sessions. The training material highlights the basic features and functionalities of Amira, Avizo and PerGeos Software.
    Amira-Avizo-advanced-training_1160x600
    Advanced training
    Maximize your investment and reduce your time-to-results with this advanced training specifically designed for existing users of Amira, Avizo and PerGeos Software.
    The course consists of a lecture with hands-on sessions. The training material highlights advanced features and functionalities of Amira, Avizo and PerGeos Software.
    Amira-Avizo-Custom-Dev_1160x600
    Custom development
    With over 25 years of experience in 3D and image processing and hundreds of custom projects delivered to organizations small and large, Thermo Fisher Scientific can provide you with a solution tailored to fit your specific needs.
    We can customize and expand our software solutions at various levels.
    Testimonials
    Video Player is loading.
    Current Time 0:00
    Duration 2:26
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 2:26
    1x
      • Chapters
      • descriptions off, selected
      • en (Main), selected
      Dr. Antentor Hinton Jr, has made extensive use of Amira Software as part of his lab’s work in mitochondrial research.
      Amira Software is crucial for visualizing SBF-SEM (serial block-face SEM) and FIB-SEM images, so we can analyze the 3D morphology of organelles.
      Dr. Antentor Hinton Jr
      Department of Molecular Physiology and Biophysics, Vanderbilt University
      Video Player is loading.
      Current Time 0:00
      Duration 6:00
      Loaded: 0.00%
      Stream Type LIVE
      Remaining Time 6:00
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Matthia A. Karreman at EMBL and DKFZ, Heidelberg, Germany, uses Amira Software
        This testimonial highlights the way Amira Software supported Matthia A. Karreman, MSc, PhD at EMBL and DKFZ, Heidelberg, Germany, in her project on multimodal correlative microscopy to efficiently target single tumor cells in vivo.
        Matthia A. Karreman, MSc, PhD
        EMBL and DKFZ, Heidelberg, Germany
        Heidelberg University uses Amira Software
        "Amira Software helps to set up complex data processing workflows. The first aim was to enhance the contrast of viral and cellular membranes. We applied a combination of 3D non-local means filter and the Membrane Enhancement Filter implemented in Amira Software. An initial automated segmentation was achieved using the Top-hat segmentation tool. This initial segmentation was further manually refined utilizing a Wacom graphics tablet. Based on the refined membrane segmentation, a 3D surface was calculated using unconstrained smoothing, and the surface was visualized as 3D model."
        By Steffen Klein (Chlanda lab, Department of Infectious Diseases, Virology, Heidelberg University, Heidelberg, Germany)
        Read more
        Resources
        Video Player is loading.
        Current Time 0:00
        Duration 2:00
        Loaded: 8.32%
        Stream Type LIVE
        Remaining Time 2:00
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Amira software solution for the analysis of time series data of cellular processes.
          Video Player is loading.
          Current Time 0:00
          Duration 1:04
          Loaded: 15.54%
          Stream Type LIVE
          Remaining Time 1:04
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • default, selected
            Amira Software can automatically count the cell nuclei which correspond to the cell count of the 3D spheroid cell cultures.
            Video Player is loading.
            Current Time 0:00
            Duration 1:58
            Loaded: 0.00%
            Stream Type LIVE
            Remaining Time 1:58
            1x
              • Chapters
              • descriptions off, selected
              • captions off, selected
              • en (Main), selected
              With incredible speed and flexibility, Amira Software enables advanced 2D-5D bioimaging workflows in research areas ranging from structural and cellular biology, to spheroid or tumor analysis and tissue imaging.
              Video Player is loading.
              Current Time 0:00
              Duration 1:57
              Loaded: 8.49%
              Stream Type LIVE
              Remaining Time 1:57
              1x
                • Chapters
                • descriptions off, selected
                • captions off, selected
                • default, selected
                Golgi apparatus of green alga Chlamydomonas reihardtii, flas-frozen without any artificial staining or fixatives, and thinned by cryo-FIB milling.
                Video Player is loading.
                Current Time 0:00
                Duration 0:30
                Loaded: 32.43%
                Stream Type LIVE
                Remaining Time 0:30
                1x
                  • Chapters
                  • descriptions off, selected
                  • captions off, selected
                  • default, selected
                  SARS-CoV-2 virion budding and assembly at the ERGIC membrane. A 3D volume rendering is shown for each area with cellular and viral membranes in green and magenta respectively, viral spike proteins in yellow, and viral ribonucleoproteins in cyan.
                  Video Player is loading.
                  Current Time 0:00
                  Duration 4:41
                  Loaded: 0.00%
                  Stream Type LIVE
                  Remaining Time 4:41
                  1x
                    • Chapters
                    • descriptions off, selected
                    • captions off, selected
                    • en (Main), selected
                    Amira Software provides a comprehensive toolset for the flexible and accurate analysis of time series data of cellular processes.
                    Video Player is loading.
                    Current Time 0:00
                    Duration 2:00
                    Loaded: 0.00%
                    Stream Type LIVE
                    Remaining Time 2:00
                    1x
                      • Chapters
                      • descriptions off, selected
                      • captions off, selected
                      • en (Main), selected
                      Amira software solution for the analysis of time series data of cellular processes.
                      Video Player is loading.
                      Current Time 0:00
                      Duration 1:04
                      Loaded: 15.54%
                      Stream Type LIVE
                      Remaining Time 1:04
                      1x
                        • Chapters
                        • descriptions off, selected
                        • captions off, selected
                        • default, selected
                        Amira Software can automatically count the cell nuclei which correspond to the cell count of the 3D spheroid cell cultures.
                        Video Player is loading.
                        Current Time 0:00
                        Duration 1:58
                        Loaded: 0.00%
                        Stream Type LIVE
                        Remaining Time 1:58
                        1x
                          • Chapters
                          • descriptions off, selected
                          • captions off, selected
                          • en (Main), selected
                          With incredible speed and flexibility, Amira Software enables advanced 2D-5D bioimaging workflows in research areas ranging from structural and cellular biology, to spheroid or tumor analysis and tissue imaging.
                          Video Player is loading.
                          Current Time 0:00
                          Duration 1:57
                          Loaded: 8.49%
                          Stream Type LIVE
                          Remaining Time 1:57
                          1x
                            • Chapters
                            • descriptions off, selected
                            • captions off, selected
                            • default, selected
                            Golgi apparatus of green alga Chlamydomonas reihardtii, flas-frozen without any artificial staining or fixatives, and thinned by cryo-FIB milling.
                            Video Player is loading.
                            Current Time 0:00
                            Duration 0:30
                            Loaded: 0.00%
                            Stream Type LIVE
                            Remaining Time 0:30
                            1x
                              • Chapters
                              • descriptions off, selected
                              • captions off, selected
                              • default, selected
                              SARS-CoV-2 virion budding and assembly at the ERGIC membrane. A 3D volume rendering is shown for each area with cellular and viral membranes in green and magenta respectively, viral spike proteins in yellow, and viral ribonucleoproteins in cyan.
                              Video Player is loading.
                              Current Time 0:00
                              Duration 4:41
                              Loaded: 0.00%
                              Stream Type LIVE
                              Remaining Time 4:41
                              1x
                                • Chapters
                                • descriptions off, selected
                                • captions off, selected
                                • en (Main), selected
                                Amira Software provides a comprehensive toolset for the flexible and accurate analysis of time series data of cellular processes.
                                Features
                                Import and process your imaging data
                                  • Handle any modality, at any scale, of any size:
                                  - Bio-Formats
                                  - Bitmap formats
                                  - Microscopy: electron and optical
                                  - Medical and neuroimage formats
                                  - Molecular formats
                                  - Other acquisition devices (MRI, radiography, etc.)
                                  • Finite element modeling, geometric modeling, CAD
                                  • Support for multi-data/multi-view, multi-channel, time series, very large data
                                  • Scaling, calibration, conversion, re-sampling
                                  • Image enhancement, comprehensive filtering and convolution, Fourier frequency transforms
                                  • Artifact reduction algorithms
                                  • Advanced multi-mode 2D/3D automatic registration
                                  • Image stack alignment, arithmetic, correlation, fusion
                                  Easily segment your imaging data
                                  • Thresholding and auto-segmentation, object separation, automatic labeling
                                  • Region growing, snakes, interpolation, wrapping, smoothing
                                  • Morphological processing, including watershed and basins
                                  • Machine Learning-based segmentation
                                  • Automatic tracing of individual fibers and filaments
                                  • Skeletonization and filament network extraction
                                  • Interactive tools for generation or editing of segmentation and spatial graphs
                                  • 3D surface reconstruction
                                  • Grid generation for FEA/CFD
                                  Export your analysis and visualization work to seamlessly publish and present it
                                  • Animation and video generation
                                  • Advanced key frame and object animation
                                  • Mix images, geometric models, measurements and simulations
                                  • Annotations, measures legends, histograms and curve plots
                                  • Export spreadsheets, 3D models, high-quality images
                                  • Active and passive 3D stereo vision
                                  • Single and tiled screen display
                                  • Immersive environment
                                  Visualize and explore your imaging data
                                  • Interactive high-quality volume and multichannel visualization
                                  • Orthogonal, oblique, cylindrical and curved slicing
                                  • Contouring and iso-surface extraction
                                  • Maximum Intensity or other types of projections
                                  • Vector and tensor visualization
                                  • Objects and tracks
                                  • Molecular visualization
                                  Analyze your imaging data and obtain quantitative information
                                  • Intuitive recipe creation, customization, automated replay
                                  • Built-in measurements, including counts, volumes, areas, perimeters, aspect ratios and orientations
                                  • User-defined measures
                                  • Results viewer with spreadsheet tool and charting
                                  • Automatic individual feature measurements, 3D localization and spreadsheet selection
                                  • Automated statistics, distribution graphs
                                  • Feature filtering using any measurement criterion
                                  • Data registration, deformation, comparison and measurements
                                  Easily and quickly adapt Amira Software to your specific needs
                                  • Custom C++ modules development
                                  • MATLAB™ bridge
                                  • Python scripting API
                                  Contact us
                                  For Research Use Only. Not for use in diagnostic procedures.
                                  Style Sheet for Global Design System
                                  Style Sheet for Komodo Tabs
                                  Style Sheet for Support and Service footer
                                  Style Sheet for Fonts
                                  Style Sheet for Cards