KingFisher Sample Purification Systems
KingFisher instruments for sample purification
KingFisher purification instruments automate the extraction and purification of DNA, RNA, proteins, cells, and exosomes. KingFisher purification instrument models serve applications ranging from oncology biomarker analyte isolation for downstream research into cancer mutation detection, microbial nucleic acid isolation for pathogen detection, to isolating genomic DNA (gDNA) for genetic testing. Their broad functionality and customizable software serve a variety of samples and target analytes such as cell-free DNA from biofluids, circulating tumor cell (CTC) and exosome isolation from liquid biopsies, nucleic acid isolation from FFPE tissue biopsies, and protein capture and digestion from select biofluids.
KingFisher instruments provide easy-to-follow protocols that remove manual steps and reduce overall processing time and errors associated with sample preparation while increasing the yield reproducibility of your results compared to manual workflows. Our data shows that samples processed with KingFisher instruments maintain consistent downstream analysis across results runs and different users, which is crucial for more sensitive applications, such as qPCR, NGS, digital PCR, and mass spectrometry.
Video Player is loading.
Current Time 0:00
Duration 1:22
Loaded: 0%
Stream Type LIVE
Remaining Time 1:22
1x
    • Chapters
    • descriptions off, selected
    • en (Main), selected
    Capabilities and applications for KingFisher instruments
    Find the right kit for your research application
    Use our KingFisher Kit Selection Tool to find the right magnetic bead-based kit for your automated sample preparation
    Prepare for the future of your sample preparation
    Learn how automated sample preparation solutions can help set you up for success in your research.
    Bead-based sample preparation
    KingFisher instruments can automate the isolation of DNA, RNA, proteins, exosomes, viruses, and cells, helping ensure reproducible results and minimized hands-on time.
    Sample preparation solutions for cancer research
    Accelerate your path to discovery with a comprehensive collection of resources, tools, and solutions to inspire your cancer research.
    KingFisher PlasmidPro Maxi Processor
    Automate your plasmid purification and explore an array of plasmid DNA purification solutions to help meet your research needs
    Interested in diagnostic nucleic acid extraction?
    The KingFisher Apex Dx system is designed to be a part of a streamlined modular workflow providing precious results, seamless data management and robust security features. Pair it with MagMAX Dx Viral/Pathogen NA Isolation Kit for nucleic acid extraction from respiratory sample types.
    For Research Use Only. Not for use in diagnostic procedures.