Search Thermo Fisher Scientific
High throughput gene expression analysis is a powerful technique that allows for the simultaneous examination of thousands of genes across multiple samples, providing comprehensive insights into cellular functions and biological processes. TaqMan Arrays contain TaqMan Gene Expression Assays dried down in three array formats: 96- and 384-well TaqMan Array plates, 384-well TaqMan Array cards (microfluidic cards), and OpenArray plates. These qPCR arrays are ideal for gene expression profiling and verification applications that require the analysis of tens to thousands of targets. The streamlined workflow of these arrays, which involves simply adding master mix and cDNA samples, helps ensure efficient and reproducible results, facilitating robust data acquisition and analysis. Additionally, their flexibility in format and customizable panel options make them suitable for a wide range of research needs, from small-scale studies to large-scale genomic projects.
These products feature our most popular TaqMan gene expression assays targeting relevant genes for specific biological pathways, processes, or diseases. Use the TaqMan Assay search tool to search for TaqMan Array cards or plates or browse the complete list of inventoried TaqMan arrays.
What chromosome position are you interested in?
Genome Version: Build GRCh38 for
For scientists who prefer not to design their panels from scratch, flexible content TaqMan arrays are an excellent choice. Users can begin by selecting a panel of commonly studied pathways, diseases, biological processes, or cellular functions (human, mouse, or rat). Each panel is pre-populated with specifically chosen assays and serves as a starting point for modification using a simple online configuration tool found on the webpage linked below. Within the tool, users can swap out genes of interest, reposition assays, or change the layout to fit their experimental design. The minimum order quantity is six plates or ten cards.
Explore:Flexible TaqMan arrays
Customized cards and plates allow researchers to configure their panel from scratch by choosing from any of the 2.8 million predesigned TaqMan Gene Expression assays. By uploading their assay list, the simple-to-use online configuration tool facilitates easy drag-and-drop arrangement of assays. The minimum order quantity is six plates or ten cards.
Learn more:Custom TaqMan array plate (96-well 0.1 mL or 0.2 mL)
Learn more:Custom TaqMan array card (384-well)
Learn more:Custom OpenArray plate
For a custom TaqMan Array that incorporates custom TaqMan gene expression assays, TaqMan SNP genotyping assays, TaqMan copy number assays, TaqMan miRNA assays, or a combination of these assay types, the Specialty TaqMan plating service is available. This service provides TaqMan assays in 96- or 384-well plates, as well as 384-well microfluidic cards, all configured to meet specific research needs.
Learn more:Specialty TaqMan array cards and plates
Product format/Instrument | QuantStudio 3 | QuantStudio 5 | QuantStudio 12K Flex |
96-well plate, standard standard 20 µL | ✓* | ✓** | ✓ |
96-well plate, fast 10 µL | ✓* | ✓** | ✓ |
384-well plate | ✓** | ✓ | |
TaqMan array card (TAC) | ✓ | ||
OpenArray | ✓ |
*QuantStudio 3 comes with either a 96-well standard OR 96-well fast block
**QuantStudio 5 comes with either a 96-well standard, 96-well fast block, OR 384-well block