Sensitivity compared to fluorescent assay detection with MUNANA substrate
Dilutions of Influenza Type B (ATCC VR-1535) virus culture supernatant (cultured on MDCK cells) were assayed at different temperatures and signal/noise (S/N) ratio calculated using uninfected MDCK cell supernatant (noise). The lower limit of detection (S/N = 2) is at least 30-fold lower with the chemiluminescent NA-Star™ assay, and S/N is approximately 50-fold higher with NA-Star assay. The dynamic range of detection with NA-Star assay with virus samples is three orders of magnitude.
Dilutions of Influenza Type B (ATCC VR-1535) virus culture supernatant (cultured on MDCK cells) were assayed at different temperatures and signal/noise (S/N) ratio calculated using uninfected MDCK cell supernatant (noise). The lower limit of detection (S/N = 2) is at least 30-fold lower with the chemiluminescent NA-<i>Star</i>™ assay, and S/N is approximately 50-fold higher with NA-<i>Star</i> assay. The dynamic range of detection with NA-<i>Star</i> assay with virus samples is three orders of magnitude.
Related Products
Catalog # Name Size Price (USD) Qty
4374348 NA-Star™ Influenza Neuraminidase Inhibitor Resistance Detection Reagent Set Each
1,854.65
Online Exclusive
2,000.00
Save 145.35 (7%)
4374422 NA-Star™ Influenza Neuraminidase Inhibitor Resistance Detection Kit Each
2,000.00
Related Images