HPLC and UHPLC Related Products
Maximize performance using optimized software, tubing, HPLC columns and more.
Easy, reliable, and innovative products
Thermo Scientific products for HPLC and UHPLC are designed to keep your research moving forward seamlessly. Our innovative and user-friendly software, Thermo Scientific Viper and nanoViper Fingertight Fittings, tubing, HPLC columns, and other consumables enable you to solve your chromatographic analysis challenges.
Build your ideal HPLC or UHPLC with our step-by-step configurator
Software
From software for the workstation to enterprise environments and for single to multichannel LC applications, we offer a range of products to meet your needs.
Chromeleon CDS
An integral part of your laboratory's IT structure, the Thermo Scientific Chromeleon Chromatography Data System (CDS) is vital for compliant, efficient, and reliable handling of today’s chromatography and mass spectrometry data.
Standard Instrument 
Integration (SII)
The SII plugin is based on the Chromeleon CDS and enables Thermo Scientific chromatography systems to be operated and controlled by Waters™ Empower™ 3 CDS or Agilent™ OpenLab™ software.
Aria MX
Manages all aspects of the multichannel LC system and controls systems with up to 8 pumps, 2 autosamplers and 4 independent flow paths for maximum throughput.
Ardia Platform
The Thermo Scientific Ardia Platform is a scalable software solution for modern environments. It connects and organizes projects, people, and instruments, enabling scientists to collaborate more efficiently and gain new insights faster.
Tubing, kits, and accessories
Viper and nanoViper Fingertight Fittings, tubing, kits, and accessories allow you to quickly connect HPLC modules, valves, and columns. They enable leak-free connections that improve chromatographic results by minimizing dead volume.  
Viper and nanoViper fittings require no tools for installation, fit narrow connections (e.g., 10-port valves), and combine simplicity with high performance.
Explore HPLC & UHPLC Tubing, Kits, and Accessories
HPLC and UHPLC columns
Need a reliable HPLC column? With over 40 years as a leader in HPLC and UHPLC column technology, you can rely on the quality of Thermo Scientific LC columns. Our extensive family of products offers a variety of particle sizes and column designs to meet all separation needs, including improved resolution, enhanced sensitivity, faster analysis and consistent performance. Whether you are running nano-flow separations, looking for analytes on the analytical scale, or scaling up your separations to semi-prep/preparative flow, our HPLC columns can meet all your chromatographic needs. 
Some of our most popular products target:
C18
 Aromatic  Polar  Biomolecules
Description
Reversed phase C18 chromatography columns offer high resolution and throughput for users separating mildly hydrophobic samples. Find the best phase for your application needs for HPLC or UHPLC, choosing from our wide selection of solid core and fully porous options.
Aromatic selectivity columns offer enhanced resolution when your analytes contain rings or double bonds. The unique selectivity of our biphenyl, PFP, and phenyl columns provide chromatographers the flexibility to separate sample mixes based on hydrophobicity and pi-pi interactions.
Columns with polar selectivity provide greater hydrophobic retention compared to conventional C18 columns. They allow users to run 100% aqueous mobile phases. Polar embedded, mixed-mode, and HILIC phases provide additional hydrophobic retention and resolution for small, polar analytes.
Reversed phase, size exclusion, hydrophobic interaction, and ion exchange chemistries are designed and optimized for long lasting, high resolution bio-separations for complex mixtures such as proteins, monoclonal antibodies and oligonucleotides.
Autosampler vials & well plates
We provide a broad selection of sample handling and containment solutions for all your chromatography requirements. Our vials, closures and well plates come with the lowest levels of extractables and leachables, are made from glass that has low compound adsorption, and the highest standards and level of certification available in the marketplace. 
Whether you have routine and robust samples, or you need to ensure the highest level of confidence and compliance, our market-leading portfolio of storage and autosampler vials and closures, well plates and sample handling accessories has everything you need. 
Vial kits
Vials & vial caps
Autosampler well plates and mats
Description
Suitable for HPLC and UHPLC, our vial kits save time during sample preparation and reduce the risk of contamination. Each kit contains 100 vials, caps and septa. They are suitable for most application requirements.
Our 2 mL vials and vial caps are available to purchase separately or as part of a kit. Available in 2 mL or low-volume options, the vials feature the lowest levels of extractables and leachables.
Our WebSeal well plates and mats are compatible with HPLC autosamplers and are available in plastic, glass or glass-coated options.
Sample preparation
Our solutions include SPE, SLE, protein precipitation, QuEChERs, filtration, protein digestion technologies and automated platforms to enable fast, reproducible and easy sample preparation for all workflows. From biopharmaceutical and pharmaceutical analysis, clinical research and toxicology studies, to food and environmental testing, Thermo Scientific sample preparation products deliver the results you need.
HPLC standards, reagents & solvents
Control retention time and selectivity, resolve complex ionic mixtures without using ion exchange columns and improve peak symmetry for your chromatography analyses by using the right reagents.
HPLC, Thermo grade solvents, clear bottle, 418x235
Contact us to get a quote or discuss an HPLC/UHPLC solution