Learn more about optical emission spectrometry
An industry-standard analytical solution for metallurgical applications, optical emission spectrometry is well-established for the analysis of a large range of metals and alloys (from Ag to Zn). It is the ideal technology for process and quality control in metal production and is routinely applied directly in-the-field for foundry production. Optical emission spectrometry offers the possibility to analyze elemental composition of alloys down to the trace level with minimal impact on operating throughput, ensuring precise and reliable readings for up to 40 individual alloying elements in a single minute. It is also possible to investigate non-metallic inclusions using advanced optical emission spectrometry methods.
If you would like to learn more about using optical emission spectrometry in your application area, speak with one of our expert sales personnel.
Demonstration center
Take advantage of a customized virtual demo experience to show you how Thermo Scientific optical emission spectrometry instruments can help with your metal process control.
Optical emission spectrometry video library
Optical emission spectrometry webinars